EMAIL THIS PAGE TO A FRIEND
The Analyst
Interdependence of initial cell density, drug concentration and exposure time revealed by real-time impedance spectroscopic cytotoxicity assay.
PMID 25868456
Abstract
We investigated the combined effect of the initial cell density (12,500, 35,000, 75,000, and 100,000 cells cm(-2)) and concentration of the anti-cancer drug doxorubicin on HeLa cells by performing time-dependent cytotoxicity assays using real-time electrochemical impedance spectroscopy. A correlation between the rate of cell death and the initial cell seeding density was found at 2.5 μM doxorubicin concentration, whereas this was not observed at 5 or 100 μM. By sensing the changes in the cell-substrate interaction using impedance spectroscopy under static conditions, the onset of cytotoxicity was observed 5 h earlier than when using a standard colorimetric end-point assay (MTS) which measures changes in the mitochondrial metabolism. Furthermore, with the MTS assay no cytotoxicity was observed after 15 h of incubation with 2.5 μM doxorubicin, whereas the impedance showed at this time point cell viability that was below 25%. These results indicate that impedance detection reveals cytotoxic events undetectable when using the MTS assay, highlighting the importance of combining impedance detection with traditional drug toxicity assays towards a more in depth understanding of the effect of anti-cancer drugs on in vitro assays. Moreover, the detection of doxorubicin induced toxicity determined with impedance under static conditions proved to be 6 times faster than in perfusion culture.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85060701
HeLa B