EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Azole antifungal inhibition of buprenorphine, methadone and oxycodone in vitro metabolism.
PMID 25868557
Abstract
Opioid-related mortality rates have escalated. Drug interactions may increase blood concentrations of the opioid. We therefore used human liver microsomes (HLMs) and cDNA-expressed human cytochrome P450s (rCYPs) to study in vitro inhibition of buprenorphine metabolism to norbuprenorphine (CYP3A4 and 2C8), oxycodone metabolism to noroxycodone (CYP3A4 and 2C18) and oxymorphone (CYP2D6), and methadone metabolism to R- and S-2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP; CYP3A4 and 2B6). In this study, we have examined the inhibitory effect of 12 (mostly antifungal) azoles. These compounds have a wide range of solubility; to keep organic solvent ≤1%, there was an equally wide range of highest concentration tested (e.g., itraconazole 5 µM to fluconazole 1000 µM). Inhibitors were first incubated with HLMs at three concentrations with or without preincubation of inhibitor with reducing equivalents to also screen for time-dependent inhibition (TDI). Posaconazole displayed evidence of TDI; metronidazole and albendazole had no significant effect. Azoles were next screened at the highest achievable concentration for non-CYP3A4 pathways. IC50 values (µM) were determined for most CYP3A4 pathways (ranges) and other pathways as dictated by screen results: clotrimazole (0.30 - 0.35; others >30 µM); econazole (2.2 - 4.9; 2B6 R-EDDP - 9.5, S-EDDP - 6.8; 2C8 - 6.0; 2C18 - 1.0; 2D6 - 1.2); fluconazole (7.7 - 66; 2B6 - 313, 361; 2C8 - 1240; 2C18 - 17; 2D6 - 1000); itraconazole (2.5 to >5; others >5); ketoconazole (0.032 - 0.094; 2B6 - 12, 31; 2C8 - 78; 2C18 - 0.98; 2D6 - 182); miconazole (2.3 - 7.6; 2B6 - 2.8, 2.8; 2C8 - 5.3; 2C18 - 3.1; 2D6 - 5.9); posaconazole (3.4 - 20; 2C18 - 3.8; others >30); terconazole (0.48 to >10; 2C18 - 8.1; others >10) and voriconazole (0.40 - 15; 2B6 - 2.4, 2.5; 2C8 - 170; 2C18 - 13; 2D6 >300). Modeling based on estimated Ki values and plasma concentrations from the literature suggest that the orally administered azoles, particularly ketoconazole and voriconazole, have the greatest potential for inhibiting CYP3A4 pathways, as does voriconazole for the CYP2B6 pathways. Azoles used for mucosal and topical applications did not exceed the modeling threshold.