EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
c-Src/Pyk2/EGFR/PI3K/Akt/CREB-activated pathway contributes to human cardiomyocyte hypertrophy: Role of COX-2 induction.
PMID 25869400
Abstract
Thrombin and COX-2 regulating cardiac hypertrophy are via various signaling cascades. Several transcriptional factors including CREB involve in COX-2 expression. However, the interplay among thrombin, CREB, and COX-2 in primary human neonatal ventricular cardiomyocytes remains unclear. In this study, thrombin-induced COX-2 promoter activity, mRNA and protein expression, and PGE2 synthesis were attenuated by pretreatment with the inhibitors of c-Src (PP1), Pyk2 (PF431396), EGFR (AG1478), PI3K/Akt (LY294002/SH-5), and p300 (GR343), or transfection with siRNAs of c-Src, Pyk2, EGFR, p110, Akt, CREB, and p300. Moreover, thrombin-stimulated phosphorylation of c-Src, Pyk2, EGFR, Akt, CREB and p300 was attenuated by their respective inhibitors. These results indicate that thrombin-induced COX-2 expression is mediated through PAR-1/c-Src/Pyk2/EGFR/PI3K/Akt linking to CREB and p300 cascades. Functionally, thrombin-induced hypertrophy and ANF/BNP release were, at least in part, mediated through a PAR-1/COX-2-dependent pathway. We uncover the importance of COX-2 regarding human cardiomyocyte hypertrophy that will provide a therapeutic intervention in cardiovascular diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503952
Anti-phospho-p300 (pSer89) antibody produced in rabbit, affinity isolated antibody
SAB4300173
Anti-phospho-PTK2B (pTyr402) antibody produced in rabbit, affinity isolated antibody
P6614 Anti-phospho-Pyk2 (pTyr402) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504522
Anti-phospho-PYK2 (pTyr402) antibody produced in rabbit, affinity isolated antibody
SAB4504678
Anti-phospho-PYK2 (pTyr579) antibody produced in rabbit, affinity isolated antibody
SAB4503831
Anti-phospho-PYK2 (pTyr580) antibody produced in rabbit, affinity isolated antibody
SAB4503830
Anti-phospho-PYK2 (pTyr881) antibody produced in rabbit, affinity isolated antibody
SAB4504644
Anti-phospho-Src (pSer75) antibody produced in rabbit, affinity isolated antibody
SAB4504669
Anti-phospho-Src (pTyr216) antibody produced in rabbit, affinity isolated antibody
S1940 Anti-phospho-Src (pTyr418) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504536
Anti-phospho-Src (pTyr418) antibody produced in rabbit, affinity isolated antibody
S2065 Anti-phospho-Src (pTyr529) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504537
Anti-phospho-Src (pTyr529) antibody produced in rabbit, affinity isolated antibody
SAB4300126
Anti-phospho-SRC (pTyr529) antibody produced in rabbit, affinity isolated antibody
SAB4300468
Anti-PTK2B (Ab-402) antibody produced in rabbit, affinity isolated antibody
AV32685
Anti-PTK2B (AB1) antibody produced in rabbit, affinity isolated antibody
AV32684
Anti-PTK2B (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1405790
Anti-PTK2B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA026091
Anti-PTK2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA026276
Anti-PTK2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
P3902 Anti-Pyk2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500835
Anti-PYK2 antibody produced in rabbit, affinity isolated antibody
SAB4500836
Anti-PYK2 antibody produced in rabbit, affinity isolated antibody
SAB4500837
Anti-PYK2 antibody produced in rabbit, affinity isolated antibody
SAB4500834
Anti-PYK2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502844
Anti-SRC, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502845
Anti-SRC, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1300875
Anti-SRC1 (C-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1307120
ANTI-SRC1(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
RAB0386 Brain Natriuretic Peptide EIA Kit, for serum, plasma, culture supernatant and cell lysates