EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
TAGLN2 regulates T cell activation by stabilizing the actin cytoskeleton at the immunological synapse.
PMID 25869671
Abstract
The formation of an immunological synapse (IS) requires tight regulation of actin dynamics by many actin polymerizing/depolymerizing proteins. However, the significance of actin stabilization at the IS remains largely unknown. In this paper, we identify a novel function of TAGLN2--an actin-binding protein predominantly expressed in T cells--in stabilizing cortical F-actin, thereby maintaining F-actin contents at the IS and acquiring LFA-1 (leukocyte function-associated antigen-1) activation after T cell receptor stimulation. TAGLN2 blocks actin depolymerization and competes with cofilin both in vitro and in vivo. Knockout of TAGLN2 (TAGLN2(-/-)) reduced F-actin content and destabilized F-actin ring formation, resulting in decreased cell adhesion and spreading. TAGLN2(-/-) T cells displayed weakened cytokine production and cytotoxic effector function. These findings reveal a novel function of TAGLN2 in enhancing T cell responses by controlling actin stability at the IS.