EMAIL THIS PAGE TO A FRIEND
Nature communications
Honokiol blocks and reverses cardiac hypertrophy in mice by activating mitochondrial Sirt3.
PMID 25871545
Abstract
Honokiol (HKL) is a natural biphenolic compound derived from the bark of magnolia trees with anti-inflammatory, anti-oxidative, anti-tumour and neuroprotective properties. Here we show that HKL blocks agonist-induced and pressure overload-mediated, cardiac hypertrophic responses, and ameliorates pre-existing cardiac hypertrophy, in mice. Our data suggest that the anti-hypertrophic effects of HKL depend on activation of the deacetylase Sirt3. We demonstrate that HKL is present in mitochondria, enhances Sirt3 expression nearly twofold and suggest that HKL may bind to Sirt3 to further increase its activity. Increased Sirt3 activity is associated with reduced acetylation of mitochondrial Sirt3 substrates, MnSOD and oligomycin-sensitivity conferring protein (OSCP). HKL-treatment increases mitochondrial rate of oxygen consumption and reduces ROS synthesis in wild type, but not in Sirt3-KO cells. Moreover, HKL-treatment blocks cardiac fibroblast proliferation and differentiation to myofibroblasts in a Sirt3-dependent manner. These results suggest that HKL is a pharmacological activator of Sirt3 capable of blocking, and even reversing, the cardiac hypertrophic response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5313
Anti-Sir2 (AS-16) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S5322
Anti-Sirt1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500493
Anti-SIRT1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501994
Anti-SIRT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
S5447
Anti-SIRT1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA006295
Anti-SIRT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32386
Anti-SIRT1 antibody produced in rabbit, affinity isolated antibody
561932
Deblock trichloroacetic acid solution, 3 % (w/v) in methylene chloride
C2HCl3O2
L7875
DL-Leucine, ≥99% (HPLC)
C6H13NO2
S5196
Monoclonal Anti-Sirt1 antibody produced in mouse, ~2 mg/mL, clone SIR11, purified immunoglobulin, buffered aqueous solution
WH0023411M1
Monoclonal Anti-SIRT1 antibody produced in mouse, clone 7B7, purified immunoglobulin, buffered aqueous solution
SAB1404972
Monoclonal Anti-SIRT1, (C-terminal) antibody produced in mouse, clone 7C2, purified immunoglobulin, buffered aqueous solution
PLA0132
Rabbit anti-Sirt1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
672920
TCA Deblock, 3 % (w/v) in methylene chloride
C2HCl3O2
T4885
Trichloroacetic acid, ≥99.0% (titration)
C2HCl3O2
T8657
Trichloroacetic acid, for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
C2HCl3O2
T0324
Trichloroacetic acid solution, ~6.25 % (w/v), 0.38 N
C2HCl3O2