EMAIL THIS PAGE TO A FRIEND
Plant, cell & environment
Tobacco guard cells fix CO2 by both Rubisco and PEPcase while sucrose acts as a substrate during light-induced stomatal opening.
PMID 25871738
Abstract
Transcriptomic and proteomic studies have improved our knowledge of guard cell function; however, metabolic changes in guard cells remain relatively poorly understood. Here we analysed metabolic changes in guard cell-enriched epidermal fragments from tobacco during light-induced stomatal opening. Increases in sucrose, glucose and fructose were observed during light-induced stomatal opening in the presence of sucrose in the medium while no changes in starch were observed, suggesting that the elevated fructose and glucose levels were a consequence of sucrose rather than starch breakdown. Conversely, reduction in sucrose was observed during light- plus potassium-induced stomatal opening. Concomitant with the decrease in sucrose, we observed an increase in the level as well as in the (13) C enrichment in metabolites of, or associated with, the tricarboxylic acid cycle following incubation of the guard cell-enriched preparations in (13) C-labelled bicarbonate. Collectively, the results obtained support the hypothesis that sucrose is catabolized within guard cells in order to provide carbon skeletons for organic acid production. Furthermore, they provide a qualitative demonstration that CO2 fixation occurs both via ribulose-1,5-biphosphate carboxylase/oxygenase (Rubisco) and phosphoenolpyruvate carboxylase (PEPcase). The combined data are discussed with respect to current models of guard cell metabolism and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
33214
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
C4H6O3
A6404
Acetic anhydride, reagent grade, ≥98%
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
L095004
Acetic anhydride
C4H6O3
M9546
D-Mannitol, BioXtra, ≥98% (GC)
C6H14O6
M9647
D-Mannitol, ACS reagent
C6H14O6
M4125
D-Mannitol, ≥98%
C6H14O6
M1902
D-Mannitol, plant cell culture tested
C6H14O6
M8429
D-Mannitol, meets EP, FCC, USP testing specifications
C6H14O6
63559
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C6H14O6
33045-U
Methoxyamine hydrochloride, bottle of 5 g
CH5NO · HCl
226904
Methoxyamine hydrochloride, 98%
CH5NO · HCl
225517
Methoxylamine hydrochloride solution, 25-30 wt. % in H2O
CH5NO · HCl
M-132
MSTFA Reagent, ampule of 10 × 1.2 mL, analytical standard
C6H12F3NOSi
M0789
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥97.0% (GC)
C5H3F6NO2
M9513
N-Methyl-bis(trifluoroacetamide), ~95% (capillary GC)
C5H3F6NO2
M7891
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, suitable for silylation
C6H12F3NOSi
394866
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
C6H12F3NOSi
270970
Pyridine, anhydrous, 99.8%
C5H5N
184527
Pyridine, spectrophotometric grade, ≥99%
C5H5N
W296600
Pyridine, ≥99%
C5H5N
24-6510
Pyridine, SAJ first grade, ≥99.0%
C5H5N
L093000
Pyridine
C5H5N
48305-U Pyridine solution, certified reference material, 2000 μg/mL in methanol