EMAIL THIS PAGE TO A FRIEND
Experimental and molecular pathology
Effect of carvedilol against myocardial injury due to ischemia-reperfusion of the brain in rats.
PMID 25872160
Abstract
We have previously reported the mechanism behind the myocardial injury and the activation of autonomic nervous system during the ischemia-reperfusion (IR) of the rat brain. This study was planned to investigate the effect of carvedilol, a β-blocker, in improving the myocardial injury caused by IR of the rat brain. We have used a whole cerebral IR model in rats by clamping both the right and left common carotid arteries. Rats were divided into five groups; Sham surgery group (Group-Sham), carvedilol treatment before ischemia group (Group-Is+C), vehicle control group (Group-Is+V), carvedilol treatment before reperfusion group (Group-Re+C) and the vehicle control group (Group-Re+V). We have measured the blood pressure and heart rate via a catheter, myocardial tissue β1-adrenaline receptor (β1-AR) levels, phosphor-p38 mitogen-activated protein kinase (p-p38 MAPK) signaling factor, malondialdehyde (MDA), and apoptosis (TUNEL assay and expression of caspase-7 protein). The results indicated that the increased expressions of β1-AR, p-p38 MAPK, caspase-7, apoptotic cells and MDA level in the myocardial tissue due to brain ischemia-reperfusion were significantly reduced by carvedilol treatment. From these observations we can suggest that, with the advantage of its antioxidant and β blocking action, carvedilol had played the improvement of myocardial injury in ischemia-reperfusion of the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4388
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
C2HBrClF3