EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Bioaccumulative characteristics of tetrabromobisphenol A and hexabromocyclododecanes in multi-tissues of prey and predator fish from an e-waste site, South China.
PMID 25874430
Abstract
Tetrabromobisphenol A (TBBPA) and hexabromocyclododecanes (HBCDs) were analyzed in 12 tissues of prey (mud carp) and predator (northern snakehead) fish from an e-waste area, South China. The TBBPA concentrations in different tissues ranged from 0.03 to 2.85 ng/g wet weight (ww) in mud carp and 0.04 to 1.30 ng/g ww in northern snakehead. The concentrations of HBCDs ranged from 0.07 to 96.9 ng/g ww in mud carp and 0.18 to 240 ng/g ww in northern snakehead. HBCD levels in tissues were correlated with lipid content for both fish species, while this correlation was only found in mud carp for TBBPA. Meanwhile, northern snakehead exhibited higher HBCD levels but lower TBBPA levels than mud carps. These observations are attributed to the more polar and reactive properties of TBBPA than HBCDs. α-HBCD was the predominant diastereoisomer of HBCDs in all tissues of mud carp and northern snakehead, except for chyme of mud carp. All the analyzed tissues in mud carp showed an enrichment of (+)-α-HBCD enantiomer with EF (enantiomeric fraction) values of 0.53-0.62, but that in northern snakehead showed an enrichment of (-)-α-HBCD enantiomer with EF values of 0.35-0.5. Considering the fact that the mud carp is one of the diet items of northern snakehead, the different enantiomer accumulation characteristics of α-HBCD between the two fish species in the present study indicated that prey and predator fish could prefer to biotransform different enantiomers of α-HBCD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330396
3,3′,5,5′-Tetrabromobisphenol A, 97%
C15H12Br4O2