EMAIL THIS PAGE TO A FRIEND
Kidney international
Macrophage and epithelial cell H-ferritin expression regulates renal inflammation.
PMID 25874599
Abstract
Inflammation culminating in fibrosis contributes to progressive kidney disease. Cross-talk between the tubular epithelium and interstitial cells regulates inflammation by a coordinated release of cytokines and chemokines. Here we studied the role of heme oxygenase-1 (HO-1) and the heavy subunit of ferritin (FtH) in macrophage polarization and renal inflammation. Deficiency in HO-1 was associated with increased FtH expression, accumulation of macrophages with a dysregulated polarization profile, and increased fibrosis following unilateral ureteral obstruction in mice: a model of renal inflammation and fibrosis. Macrophage polarization in vitro was predominantly dependent on FtH expression in isolated bone marrow-derived mouse monocytes. Using transgenic mice with conditional deletion of FtH in the proximal tubules (FtH(PT-/-)) or myeloid cells (FtH(LysM-/-)), we found that myeloid FtH deficiency did not affect polarization or accumulation of macrophages in the injured kidney compared with wild-type (FtH(+/+)) controls. However, tubular FtH deletion led to a marked increase in proinflammatory macrophages. Furthermore, injured kidneys from FtH(PT-/-) mice expressed significantly higher levels of inflammatory chemokines and fibrosis compared with kidneys from FtH(+/+) and FtH(LysM-/-) mice. Thus, there are differential effects of FtH in macrophages and epithelial cells, which underscore the critical role of FtH in tubular-macrophage cross-talk during kidney injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405949
Anti-HMOX1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101053
Anti-HMOX1 antibody produced in rabbit, affinity isolated antibody
SAB1410641
Anti-HMOX1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4502012
Anti-INOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SRP3211 IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SRP4137 IL-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
SRP6172
IL-4 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SRP3093 IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I1020 Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I3650 Interleukin-4 from rat, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)
I4269 Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I2661 Interleukin-4 human, IL-4, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), animal component free
WH0003162M1
Monoclonal Anti-HMOX1 antibody produced in mouse, clone 5C6, purified immunoglobulin, buffered aqueous solution
251682
Sodium 3-mercapto-1-propanesulfonate, technical grade, 90%
C3H7NaO3S2