PloS one
iTRAQ-based quantitative proteomic analysis on S100 calcium binding protein A2 in metastasis of laryngeal cancer.
PMID 25874882
Abstract
Laryngeal cancer is the most frequent neoplasm in the head and neck region, with the vast majority of tumors originating from squamous cells. The survival rate of patients with laryngeal cancer has not improved substantially over the past 25 years. To acquire further knowledge regarding the molecules responsible for laryngeal cancer oncogenesis and, in turn, to improve target therapy iTRAQ and mass spectrometry analysis were utilized to detect differences in protein expression from 15 paired laryngeal cancer and adjacent non-cancerous tissue samples. Using mass spectrometry analysis, the expression levels of 100 proteins in laryngeal cancer samples were distinct from the non-tumor, non-cancerous samples. Further validation of the differentially expressed proteins S100A2, KRT16, FGB and HSPB1 were carried out using quantitative real-time RT-PCR, immunoblot and immunohistochemistry. Functional analysis of one of the highly expressed proteins, S100 calcium binding protein A2 (S100A2), was performed using RNA interference. As a consequence, attenuated S100A2 expression enhanced the ability of HEp-2 cell lines to migrate and invade in vitro. Our investigation complements the current understanding of laryngeal cancer progression. Furthermore, this study supports the concept that enhanced expression of S100A2 may be a promising strategy in developing novel cancer therapeutic drugs.