PloS one
A clinical approach to brown adipose tissue in the para-aortic area of the human thorax.
PMID 25875082
Abstract
Human thoracic brown adipose tissue (BAT), composed of several subdivisions, is a well-known target organ of many clinical studies; however, the functional contribution of each part of human thoracic BAT remains unknown. The present study analyzed the significance of each part of human thoracic BAT in the association between regional distribution, cellularity, and factors involved in the functional regulation of thoracic BAT. We analyzed 1550 healthy adults who underwent medical check-ups by positron-emission tomography and computed tomography (PET-CT) imaging, 8 cadavers, and 78 autopsy cases in an observational study. We first characterized the difference between the mediastinum and the supraclavicular areas using counts of BAT detection and conditions based on PET-CT outcomes. The measurable important area was then subjected to systematic anatomical and immunohistochemical analyses using anti-uncoupling protein 1 (UCP1) antibody to characterize the cellularity in association with age and sex. In PET-CT scanning, the main site of thoracic BAT was the mediastinum rather than the supraclavicular area (P < 0.05). Systemic macroanatomy revealed that the thumb-sized BAT in the posterior mediastinal descending para-aortic area (paBAT) had feeding vessels from the posterior intercostal arteries and veins and sympathetic/parasympathetic innervation from trunks of the sympathetic and vagus nerves, respectively. Immunohistochemical analysis indicated that the paBAT exhibited immunoreactivity for tyrosine hydroxylase and vesicular acetylcholine transporter located in the pericellular nervous fibers and intracellular UCP1. The brown adipose cells of paBAT showed age-dependent decreases in UCP1 expression (P < 0.05), accompanied by a significant increase in vacuole formation, indicating fat accumulation (P < 0.05), from 10 to 37 years of age (P < 0.01). paBAT may be one of the essential sites for clinical application in BAT study because of its visible anatomy with feeding vessels and sympathetic/parasympathetic innervation functionally affected by outer condition and senescence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1568
Mannose triflate, For PET imaging, ≥98% (TLC)
C15H19F3O12S