EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Rheumatologie
The osteogenic potential of ligament fibroblasts is greater in ankylosing spondylitis patients than in patients with osteoarthritis.
PMID 25876050
Abstract
The goal of the present study was to investigate the osteogenic potential of ligament fibroblasts from patients with ankylosing spondylitis (AS). Ligament fibroblasts were isolated from tissues harvested from AS patients and patients with aseptic necrosis of the femoral head (ANFH) who had undergone total hip arthroplasty (THA). Cells were cultured in osteogenic induction medium (OIM) with or without bone morphogenetic proteinxa02 (BMP-2) for 30xa0days. During this time, the levels of alkaline phosphatase (ALP) and osteocalcin (OC) were determined as markers of osteogenesis. Ligament fibroblasts isolated from patients with AS exhibited significantly greater potential for differentiation than those isolated from patients with osteoarthritis (ANFH). These findings illuminate osteogenesis as a new pathway for studying the mechanism underlying ankylosis in AS patients. Factors which cause localized stromal remodeling at the enthesis significantly influence formation of new bone and further research is required to determine the mechanisms responsible for the osteogenic potential of enthesis cells and ligament fibroblasts in AS patients.