EMAIL THIS PAGE TO A FRIEND
Journal of virology
EFTUD2 Is a Novel Innate Immune Regulator Restricting Hepatitis C Virus Infection through the RIG-I/MDA5 Pathway.
PMID 25878102
Abstract
The elongation factor Tu GTP binding domain-containing protein 2 (EFTUD2) was identified as an anti-hepatitis C virus (HCV) host factor in our recent genome-wide small interfering RNA (siRNA) screen. In this study, we sought to further determine EFTUD2's role in HCV infection and investigate the interaction between EFTUD2 and other regulators involved in HCV innate immune (RIG-I, MDA5, TBK1, and IRF3) and JAK-STAT1 pathways. We found that HCV infection decreased the expression of EFTUD2 and the viral RNA sensors RIG-I and MDA5 in HCV-infected Huh7 and Huh7.5.1 cells and in liver tissue from in HCV-infected patients, suggesting that HCV infection downregulated EFTUD2 expression to circumvent the innate immune response. EFTUD2 inhibited HCV infection by inducing expression of the interferon (IFN)-stimulated genes (ISGs) in Huh7 cells. However, its impact on HCV infection was absent in both RIG-I knockdown Huh7 cells and RIG-I-defective Huh7.5.1 cells, indicating that the antiviral effect of EFTUD2 is dependent on RIG-I. Furthermore, EFTUD2 upregulated the expression of the RIG-I-like receptors (RLRs) RIG-I and MDA5 to enhance the innate immune response by gene splicing. Functional experiments revealed that EFTUD2-induced expression of ISGs was mediated through interaction of the EFTUD2 downstream regulators RIG-I, MDA5, TBK1, and IRF3. Interestingly, the EFTUD2-induced antiviral effect was independent of the classical IFN-induced JAK-STAT pathway. Our data demonstrate that EFTUD2 restricts HCV infection mainly through an RIG-I/MDA5-mediated, JAK-STAT-independent pathway, thereby revealing the participation of EFTUD2 as a novel innate immune regulator and suggesting a potentially targetable antiviral pathway. Innate immunity is the first line defense against HCV and determines the outcome of HCV infection. Based on a recent high-throughput whole-genome siRNA library screen revealing a network of host factors mediating antiviral effects against HCV, we identified EFTUD2 as a novel innate immune regulator against HCV in the infectious HCV cell culture model and confirmed that its expression in HCV-infected liver tissue is inversely related to HCV infection. Furthermore, we determined that EFTUD2 exerts its antiviral activity mainly through governing its downstream regulators RIG-I and MDA5 by gene splicing to activate IRF3 and induce classical ISG expression independent of the JAT-STAT signaling pathway. This study broadens our understanding of the HCV innate immune response and provides a possible new antiviral strategy targeting this novel regulator of the innate response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400461
Anti-DDX58 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2104315
Anti-DDX58 antibody produced in rabbit, affinity isolated antibody
HPA022021
Anti-EFTUD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701211
Anti-EFTUD2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503626
Anti-EFTUD2 antibody produced in rabbit, affinity isolated antibody
SAB1407860
Anti-IFIH1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002656
Anti-IFIH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501565
Anti-IRF-3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406043
Anti-IRF3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004895
Anti-IRF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV31992
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV38061
Anti-IRF3 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB3500280
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501563
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501564
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB3500356
Anti-MDA5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1100069
Anti-MX1 (116-130) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406151
Anti-MX1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101536
Anti-MX1 antibody produced in rabbit, affinity isolated antibody
SAB1401252
Anti-MX1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA030917
Anti-MX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS2351
Anti-NAK antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101667
Anti-OAS1 (ab1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2101668
Anti-OAS1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2101669
Anti-OAS1 (ab3) antibody produced in rabbit, affinity isolated antibody
HPA003657
Anti-OAS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105033
Anti-OAS1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503926
Anti-phospho-IRF-3 (pSer385) antibody produced in rabbit, affinity isolated antibody
SAB4504651
Anti-phospho-IRF-3 (pSer386) antibody produced in rabbit, affinity isolated antibody
SAB4504031
Anti-phospho-IRF-3 (pSer396) antibody produced in rabbit, affinity isolated antibody