EMAIL THIS PAGE TO A FRIEND
Oncotarget
TOPK is highly expressed in circulating tumor cells, enabling metastasis of prostate cancer.
PMID 25881543
Abstract
Circulating tumor cells (CTCs) are important for metastasis in prostate cancer. T-LAK cell-originated protein kinase (TOPK) is highly expressed in cancer cells. Herein, we established a xenograft animal model, isolated and cultured the CTCs, and found CTCs have significantly greater migratory capacity than parental cells. TOPK is more highly expressed in the CTCs than in parental cells and is also highly expressed in the metastatic nodules caused by CTCs in mice. Knocking down TOPK decreased the migration of CTCs both in vitro and in vivo. TOPK was modulated by the PI3K/PTEN and ERK pathways during the metastasis of prostate cancer. High levels of TOPK in the tumors of patients were correlated with advanced stages of prostate cancer, especially for high-risk patients of Gleason score≥8, PSA>20ng/ml. In summary, TOPK was speculated to be one of a potential marker and therapeutic target in advanced prostate cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501670
Anti-Keratin 19 antibody produced in rabbit, affinity isolated antibody
SAB2101302
Anti-KRT19 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101303
Anti-KRT19 (ab2) antibody produced in rabbit, affinity isolated antibody
HPA002465 Anti-KRT19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407709
Anti-PBK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412112
ANTI-PBK antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution
SAB1412132
ANTI-PBK antibody produced in mouse, clone 3F7, purified immunoglobulin, buffered aqueous solution
SAB1412198
ANTI-PBK antibody produced in mouse, clone 2D6, purified immunoglobulin, buffered aqueous solution
SAB1412245
ANTI-PBK antibody produced in mouse, clone 3A7, purified immunoglobulin, buffered aqueous solution
SAB1412269 ANTI-PBK antibody produced in mouse, clone 4D11, purified immunoglobulin, buffered aqueous solution
SAB1412270
ANTI-PBK antibody produced in mouse, clone 2A3, purified immunoglobulin, buffered aqueous solution
SAB1412271
ANTI-PBK antibody produced in mouse, clone 2D2, purified immunoglobulin, buffered aqueous solution
SAB1412452 ANTI-PBK antibody produced in mouse, clone 4E3, purified immunoglobulin, buffered aqueous solution
WH0055872M6
Monoclonal Anti-PBK antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution
SAB5300406
Monoclonal Anti-PBK antibody produced in mouse, clone 2C8, ascites fluid
SAB1405168
Monoclonal Anti-PBK, (N-terminal) antibody produced in mouse, clone 3A11, purified immunoglobulin, buffered aqueous solution