EMAIL THIS PAGE TO A FRIEND
Journal of the American College of Cardiology
Metabolic disturbances identified in plasma are associated with outcomes in patients with heart failure: diagnostic and prognostic value of metabolomics.
PMID 25881932
Abstract
Identification of novel biomarkers is needed to improve the diagnosis and prognosis of heart failure (HF). Metabolic disturbance is remarkable in patients with HF. This study sought to assess the diagnostic and prognostic values of metabolomics in HF. Mass spectrometry-based profiling of plasma metabolites was performed in 515 participants; the discovery phase study enrolled 51 normal control subjects and 183 HF patients, and the validation study enrolled 63 control subjects and 218 patients with stage C HF. Another independent group of 32 patients with stage C HF who recovered to New York Heart Association functional class I at 6 and 12 months was profiled as the "recovery" group. A panel of metabolites, including histidine, phenylalanine, spermidine, and phosphatidylcholine C34:4, has a diagnostic value similar to B-type natriuretic peptide (BNP). In the recovery group, the values of this panel significantly improved at 6 and 12 months. To evaluate the prognostic values, events were defined as the combined endpoints of death or HF-related re-hospitalization. A metabolite panel, which consisted of the asymmetric methylarginine/arginine ratio, butyrylcarnitine, spermidine, and the total amount of essential amino acids, provided significant prognostic values (p < 0.0001) independent of BNP and traditional risk factors. The prognostic value of the metabolite panel was better than that of BNP (area under the curve of 0.85 vs. 0.74 for BNP) and Kaplan-Meier curves (log rank: 17.5 vs. 9.95). These findings were corroborated in the validation study. Metabolomics demonstrate powerful diagnostic value in estimating HF-related metabolic disturbance. The profile of metabolites provides better prognostic value versus conventional biomarkers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1270000
Arginine, European Pharmacopoeia (EP) Reference Standard
C6H14N4O2