EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Whole-exome sequencing reveals ZNF408 as a new gene associated with autosomal recessive retinitis pigmentosa with vitreal alterations.
PMID 25882705
Abstract
Retinitis pigmentosa (RP) is a group of progressive inherited retinal dystrophies that cause visual impairment as a result of photoreceptor cell death. RP is heterogeneous, both clinically and genetically making difficult to establish precise genotype-phenotype correlations. In a Spanish family with autosomal recessive RP (arRP), homozygosity mapping and whole-exome sequencing led to the identification of a homozygous mutation (c.358_359delGT; p.Ala122Leufs*2) in the ZNF408 gene. A screening performed in 217 additional unrelated families revealed another homozygous mutation (c.1621C>T; p.Arg541Cys) in an isolated RP case. ZNF408 encodes a transcription factor that harbors 10 predicted C2H2-type fingers thought to be implicated in DNA binding. To elucidate the ZNF408 role in the retina and the pathogenesis of these mutations we have performed different functional studies. By immunohistochemical analysis in healthy human retina, we identified that ZNF408 is expressed in both cone and rod photoreceptors, in a specific type of amacrine and ganglion cells, and in retinal blood vessels. ZNF408 revealed a cytoplasmic localization and a nuclear distribution in areas corresponding with the euchromatin fraction. Immunolocalization studies showed a partial mislocalization of the p.Arg541Cys mutant protein retaining part of the WT protein in the cytoplasm. Our study demonstrates that ZNF408, previously associated with Familial Exudative Vitreoretinopathy (FEVR), is a new gene causing arRP with vitreous condensations supporting the evidence that this protein plays additional functions into the human retina.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW10742A Anti-ARR3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405487
Anti-ARR3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3501086
Anti-ARR3 antibody produced in rabbit, affinity isolated antibody
SAB1411780
ANTI-RCV1 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
SAB1411781
ANTI-RCV1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1411794
ANTI-RCV1 antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
SAB1411795
ANTI-RCV1 antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
SAB1411804
ANTI-RCV1 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1411805
ANTI-RCV1 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1411862
ANTI-RCV1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1411863
ANTI-RCV1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
SAB1411864
ANTI-RCV1 antibody produced in mouse, clone 4A8, purified immunoglobulin, buffered aqueous solution
SAB1411865
ANTI-RCV1 antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
SAB1411878
ANTI-RCV1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
SAB1411879
ANTI-RCV1 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
AV38196
Anti-RCV1 antibody produced in rabbit, affinity isolated antibody
SAB1406380
Anti-RCVRN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101971
Anti-RCVRN antibody produced in rabbit, affinity isolated antibody
SAB4200309
Anti-Recoverin (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB4502604
Anti-Recoverin antibody produced in rabbit, affinity isolated antibody
SAB1402694
Monoclonal Anti-ARR3 antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution
WH0005957M1
Monoclonal Anti-RCV1 antibody produced in mouse, clone 4C6, purified immunoglobulin, buffered aqueous solution
SAB1404306
Monoclonal Anti-RCV1, (C-terminal) antibody produced in mouse, clone 3G10, purified immunoglobulin, buffered aqueous solution