EMAIL THIS PAGE TO A FRIEND
Oncotarget
miR-221/222 induces pancreatic cancer progression through the regulation of matrix metalloproteinases.
PMID 25883224
Abstract
MicroRNAs are involved in the initiation and progression of pancreatic cancer. In this study, we showed that miR-221/222 is overexpressed in pancreatic cancer. MiR-221/222 overexpression significantly promoted pancreatic cancer cell proliferation and invasion while inhibiting apoptosis. The expression of the matrix metalloproteinases (MMPs) MMP-2 and MMP-9 was increased in miR-221/222 mimic-transfected pancreatic cancer cells. Validation experiments identified TIMP-2 as a direct target of miR-221/222. These data indicate that overexpressed miR-221/222 may play an oncogenic role in pancreatic cancer by inducing the expression of MMP-2 and MMP-9, thus leading to cancer cell invasion.