PloS one
The role of toll-like receptor 9 in chronic stress-induced apoptosis in macrophage.
PMID 25885582
Abstract
Emerging evidence implied that chronic stress has been exerting detrimental impact on immune system functions in both humans and animals. Toll-like receptors (TLRs) have been shown to play an essential role in modulating immune responses and cell survival. We have recently shown that TLR9 deficiency protects against lymphocyte apoptosis induced by chronic stress. However, the exact role of TLR9 in stress-mediated change of macrophage function remains unclear. The results of the current study showed that when BALB/c mice were treated with restraint stress (12 h daily for 2 days), the number of macrophages recruited to the peritoneal cavity was obviously increased. Results also demonstrated that the sustained effects of stress elevated cytokine IL-1β, TNF-α and IL-10 production yet diminished IFN-γ production from macrophage, which led to apoptotic cell death. However, TLR9 deficiency prevented the chronic stress-mediated accumulation of macrophages. In addition, knocking out TLR9 significantly abolished the chronic stress-induced imbalance of cytokine levels and apoptosis in macrophage. TLR9 deficiency was also found to reverse elevation of plasma IL-1β, IL-10 and IL-17 levels and decrease of plasma IFN-γ level under the condition of chronic stress. These results indicated that TLR9-mediated macrophage responses were required for chronic stress-induced immunosuppression. Further exploration showed that TLR9 deficiency prevented the increment of p38 MAPK phosphorylation and reduction of Akt/Gsk-3β phosphorylation; TLR9 deficiency also attenuated the release of mitochondrial cytochrome c into cytoplasm, caused upregulation of Bcl-2/Bax protein ratio, downregulation of cleavage of caspase-3 and PARP, as well as decreased TUNEL-positive cells in macrophage of stressed mice. Collectively, our studies demonstrated that deficiency of TLR9 maintained macrophage function by modulating macrophage accumulation and attenuating macrophage apoptosis, thus preventing immunosuppression in restraint-stressed mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501330
Anti-GSK3 antibody produced in rabbit, affinity isolated antibody
SAB4300287
Anti-GSK3B (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2100978
Anti-GSK3B antibody produced in rabbit, affinity isolated antibody
HPA028017
Anti-GSK3B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300001
Anti-phospho-GSK3B (pSer9) antibody produced in rabbit, affinity isolated antibody
SAB4300237
Anti-phospho-GSK3B (pTyr216) antibody produced in rabbit, affinity isolated antibody
GW22790 Anti-TLR9 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412111 ANTI-TLR9 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
SAB1412155
ANTI-TLR9 antibody produced in mouse, clone 3B7, purified immunoglobulin, buffered aqueous solution
SAB1412156
ANTI-TLR9 antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
SAB1412299
ANTI-TLR9 antibody produced in mouse, clone 1E4, purified immunoglobulin, buffered aqueous solution
SAB1412300
ANTI-TLR9 antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
SAB1412301
ANTI-TLR9 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412302
ANTI-TLR9 antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB3500313
Anti-TLR9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104136
Anti-TLR9, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0002932M4
Monoclonal Anti-GSK3B antibody produced in mouse, clone 1H3, purified immunoglobulin, buffered aqueous solution
SAB5300175 Monoclonal Anti-GSK3B antibody produced in mouse, clone 3D10, ascites fluid
WH0054106M3
Monoclonal Anti-TLR9 antibody produced in mouse, clone 1E8, purified immunoglobulin, buffered aqueous solution
SAB1405118
Monoclonal Anti-TLR9 antibody produced in mouse, clone 1F4, purified immunoglobulin, buffered aqueous solution