EMAIL THIS PAGE TO A FRIEND
BMC cell biology
AKT regulation of mesothelial-to-mesenchymal transition in peritoneal dialysis is modulated by Smurf2 and deubiquitinating enzyme USP4.
PMID 25885904
Abstract
Transforming growth factor-β1 (TGF-β1) plays a key role in mesothelial-to-mesenchymal transition (MMT) during peritoneal dialysis (PD). However, the role of Akt in MMT transformation in PD is not clear. In this study, we observed that the phosphorylated form of protein kinase B (Akt), termed as pAkt, was up-regulated in the peritoneum of mice undergoing PD. It was associated with thickening of the peritoneum and up-regulation of TGF-β1. Upregulation of pAkt paralleled with the increased expression of Smad ubiquitination regulatory factor 2 (Smurf2), Vimentin and fibronectin (FN), and decreased expression of mothers against decapentaplegic homolog 7 (Smad7) and Zonula Occludens protein 1(ZO-1) in mice undergoing PD treatment and in TGF-β1 induced human peritoneal mesothelial cells (HPMCs). These changes were reversed with the treatment of a PI3K/Akt inhibitor LY294002 in vivo or in cells transfected with Akt dominant-negative (Akt-DN) plasmids in vitro. Increased Smurf2 expression in HPMCs, induced by TGF-β1 was accompanied with altered expression of Transforming growth factor receptor I (TβR-I), Smad7, ZO-1, Vimentin and FN via Akt modulation. In addition, inhibition of Ubiquitin carboxyl-terminal hydrolase 4 (USP4) decreased TGF- β1-induced expression of TβR-I and reversed the altered expression of Smad7, Smurf2, ZO-1 and Vimentin. Moreover, TGF-β1 accentuated the interactions between Smurf2 and Smad7, while reduced the association between TβR-I and Smurf2. These interactions were reversed by the treatment of Akt-DN and USP4 siRNA, respectively. These data implied that Akt mediated MMT in PD via Smurf2 modulation/and or Smad7 degradation while conceivably maintaining the TβRI stability, most likely by the USP4.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317632
(Methylcyclopentadienyl)manganese(I) tricarbonyl
C9H7MnO3