EMAIL THIS PAGE TO A FRIEND
BMC microbiology
Optimization of expression conditions for a novel NZ2114-derived antimicrobial peptide-MP1102 under the control of the GAP promoter in Pichia pastoris X-33.
PMID 25887810
Abstract
The infections caused by antibiotic multidrug-resistant bacteria seriously threaten human health. To prevent and cure the infections caused by multidrug-resistant bacteria, new antimicrobial agents are required. Antimicrobial peptides are ideal therapy candidates for antibiotic-resistant pathogens. However, due to high production costs, novel methods of large-scale production are urgently needed. The novel plectasin-derived antimicrobial peptide-MP1102 gene was constitutively expressed under the control of the GAP promoter. The optimum carbon source and concentration were determined, and 4% glucose (w/v) was initially selected as the best carbon source. Six media were assayed for the improved yield of recombinant MP1102 (rMP1102). The total protein and rMP1102 yield was 100.06xa0mg/l and 42.83xa0mg/l, which was accomplished via the use of medium number 1. The peptone and yeast extract from Hongrun Baoshun (HRBS, crude industrial grade, Beijing, China) more effectively improved the total protein and the yield of rMP1102 to 280.41xa0mg/l and 120.57xa0mg/l compared to 190.26xa0mg/l and 78.01xa0mg/l that resulted from Oxoid (used in the research). Furthermore, we observed that the total protein, antimicrobial activity and rMP1102 yield from the fermentation supernatant increased from 807.42 mg/l, 384,000xa0AU/ml, and 367.59 mg/l, respectively, in pH5.0 to 1213.64xa0mg/l, 153,600xa0AU/ml and 538.17xa0mg/ml, respectively in pHxa06.5 in a 5-l fermenter. Accordingly, the productivity increased from 104464xa0AU/mg rMP1102 in pHxa05.0 to a maximum of 285412xa0AU/mg rMP1102 in pHxa06.5. Finally, the recombinant MP1102 was purified with a cation-exchange column with a yield of 376.89xa0mg/l, 96.8% purity, and a molecular weight of 4382.9xa0Da, which was consistent with its theoretical value of 4383xa0Da. It's the highest level of antimicrobial peptides expressed in Pichia pastoris using GAP promoter so far. These results provide an economical method for the high-level production of rMP1102 under the control of the GAP promoter.