EMAIL THIS PAGE TO A FRIEND
BMC microbiology
Lactobacillus reuteri I5007 modulates tight junction protein expression in IPEC-J2 cells with LPS stimulation and in newborn piglets under normal conditions.
PMID 25888437
Abstract
Tight junctions (TJs) maintain the intestinal mucosal barrier, dysfunction of which plays a vital role in the pathophysiology of a variety of gastrointestinal disorders. Previously, we have shown that L. reuteri I5007 maintained the gut epithelial barrier in newborn piglets. Here we aimed to decipher the influence of L. reuteri I5007 on tight junction (TJ) protein expression both in vivo and in vitro. We found that L. reuteri I5007 significantly increased the protein abundance of intestinal epithelial claudin-1, occludin and zonula occluden-1 (ZO-1) in newborn piglets (orally administrated with 6 × 10(9)xa0CFU of L. reuteri I5007 daily for 14xa0days). In vitro, treatment with L. reuteri I5007 alone maintained the transepithelial electrical resistance (TEER) of IPEC-J2 cells with time. In addition, IPEC-J2 cells were stimulated with 1xa0μg/mL lipopolysaccharide (LPS) for 1, 4, 8, 12 or 24xa0h, following pre-treatment with L. reuteri I5007 or its culture supernatant for 2xa0h. The results showed that LPS time-dependently induced (significantly after 4 or 8xa0h) the expression of TNF-α and IL-6, and decreased TJ proteins, which was reversed by pre-treatment of L. reuteri I5007 or its culture supernatant. L. reuteri I5007 had beneficial effects on the expression of TJ proteins in newborn piglets and the in-vitro results showed this strain had a positive effect on TEER of cells and inhibited the reduction of TJ proteins expression induced by LPS. These findings indicated L. reuteri I5007 may have potential roles in protection TJ proteins in TJ-deficient conditions.