EMAIL THIS PAGE TO A FRIEND
Malaria journal
Plasmodium vivax inhibits erythroid cell growth through altered phosphorylation of the cytoskeletal protein ezrin.
PMID 25889165
Abstract
The underlying causes of severe malarial anaemia are multifactorial. In previously reports, Plasmodium vivax was found to be able to directly inhibited erythroid cell proliferation and differentiation. The molecular mechanisms underlying the suppression of erythropoiesis by P. vivax are remarkably complex and remain unclear. In this study, a phosphoproteomic approach was performed to dissect the molecular mechanism of phosphoprotein regulation, which is involved in the inhibitory effect of parasites on erythroid cell development. This study describes the first comparative phosphoproteome analysis of growing erythroid cells (gECs), derived from human haematopoietic stem cells, exposed to lysates of infected erythrocytes (IE)/uninfected erythrocytes (UE) for 24, 48 and 72xa0h. This study utilized IMAC phosphoprotein isolation directly coupled with LC MS/MS analysis. Lysed IE significantly inhibited gEC growth at 48 and 72xa0h and cell division resulting in the accumulation of cells in G0 phase. The relative levels of forty four phosphoproteins were determined from gECs exposed to IE/UE for 24-72xa0h and compared with the media control using the label-free quantitation technique. Interestingly, the levels of three phosphoproteins: ezrin, alpha actinin-1, and Rho kinase were significantly (p < 0.05) altered. These proteins display interactions and are involved in the regulation of the cellular cytoskeleton. Particularly affected was ezrin (phosphorylated at Thr567), which is normally localized to gEC cell extension peripheral processes. Following exposure to IE, for 48-72xa0h, the ezrin signal intensity was weak or absent. This result suggests that phospho-ezrin is important for actin cytoskeleton regulation during erythroid cell growth and division. These findings suggest that parasite proteins are able to inhibit erythroid cell growth by down-regulation of ezrin phosphorylation, leading to ineffective erythropoiesis ultimately resulting in severe malarial anaemia. A better understanding of the mechanisms of ineffective erythropoiesis may be beneficial in the development of therapeutic strategies to prevent severe malarial anaemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45533
3-Indoleacetic acid, PESTANAL®, analytical standard
C10H9NO2