EMAIL THIS PAGE TO A FRIEND
Oncotarget
The oncogenic receptor ErbB2 modulates gemcitabine and irinotecan/SN-38 chemoresistance of human pancreatic cancer cells via hCNT1 transporter and multidrug-resistance associated protein MRP-2.
PMID 25890497
Abstract
Pancreatic adenocarcinoma (PDAC) is one of the most deadly cancers because of a lack of early diagnostic markers and efficient therapeutics. The fluorinated analog of deoxycytidine, gemcitabine and emerging FOLFIRINOX protocol (5-fluorouracil (5-FU), irinotecan/SN-38, oxaliplatin and leucovorin) are the main chemotherapies to treat PDAC. The ErbB2/HER2 oncogenic receptor is commonly overexpressed in PDAC. In this context, we aimed to decipher the ErbB2-mediated mechanisms of chemoresistance to the two main chemotherapy protocols used to treat PDAC.ErbB2 knocking down (KD) in CAPAN-1 and CAPAN-2 cells led to an increased sensitivity to gemcitabine and an increased resistance to irinotecan/SN-38 both in vitro and in vivo (subcutaneous xenografts) This was correlated to an increase of hCNT1 and hCNT3 transporters and ABCG2, MRP1 and MRP2 ATP-binding cassette transporters expression and resistance to cell death. We also show that MRP2 is repressed following activation of JNK, Erk1/2 and NF-κB pathways by ErbB2. Finally, in datasets of human PDAC samples, ErbB2 and MRP2 expression was conversely correlated. Altogether, we propose that ErbB2 mediates several intracellular mechanisms linked to PDAC cell chemoresistance that may represent potential targets in order to ameliorate chemotherapy response and allow stratification of patients eligible for either gemcitabine or FOLFIRINOX treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-636
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, 200 µg, Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in ChIP, ICC, IF, WB. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.
HPA004860
Anti-ABCC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2106637
Anti-ABCC2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2502079
Anti-ESR2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV31652
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2100713
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2105299
Anti-ESR2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300514
Anti-H2AFX (Ab-139) antibody produced in rabbit, affinity isolated antibody
SAB4501369
Anti-Histone H2AX , C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1409579
Anti-KLHL1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021196
Anti-KLHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101283
Anti-KLHL1 antibody produced in rabbit, affinity isolated antibody
SAB3500342
Anti-KLHL1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2106333
Anti-KLHL1 antibody produced in rabbit, affinity isolated antibody
M8316 Anti-MRP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
M3692
Anti-MRP2 antibody, Mouse monoclonal, clone CPR96, purified from hybridoma cell culture
HPA023311 Anti-SLC28A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB1305880
ANTI-SYCE1L(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PLA0294
Goat anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB1403681
Monoclonal Anti-ABCC2 antibody produced in mouse, clone 2H6, purified immunoglobulin, buffered aqueous solution
WH0002100M1
Monoclonal Anti-ESR2 antibody produced in mouse, clone 3F3, purified immunoglobulin, buffered aqueous solution
SAB2702145
Monoclonal Anti-ESR2 antibody produced in mouse
SAB2702146 Monoclonal Anti-ESR2 antibody produced in mouse
SAB2702147 Monoclonal Anti-ESR2 antibody produced in mouse
E1276 Monoclonal Anti-Estrogen Receptor β antibody produced in mouse, clone 9.88, ascites fluid
WH0003014M5 Monoclonal Anti-H2AFX antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
SAB1403902 Monoclonal Anti-H2AFX, (N-terminal) antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
SAB1403331
Monoclonal Anti-KLHL1 antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
SAB5300270
Monoclonal Anti-KLHL1 antibody produced in mouse, clone 3A8, ascites fluid
PLA0023
Rabbit anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.