EMAIL THIS PAGE TO A FRIEND
Toxicology
Sodium nitrite potentiates renal oxidative stress and injury in hemoglobin exposed guinea pigs.
PMID 25891524
Abstract
Methemoglobin-forming drugs, such as sodium nitrite (NaNO2), may exacerbate oxidative toxicity under certain chronic or acute hemolytic settings. In this study, we evaluated markers of renal oxidative stress and injury in guinea pigs exposed to extracellular hemoglobin (Hb) followed by NaNO2 at doses sufficient to simulate clinically relevant acute methemoglobinemia. NaNO2 induced rapid and extensive oxidation of plasma Hb in this model. This was accompanied by increased renal expression of the oxidative response effectors nuclear factor erythroid 2-derived-factor 2 (Nrf-2) and heme oxygenase-1 (HO-1), elevated non-heme iron deposition, lipid peroxidation, interstitial inflammatory cell activation, increased expression of tubular injury markers kidney injury-1 marker (KIM-1) and liver-fatty acid binding protein (L-FABP), podocyte injury, and cell death. Importantly, these indicators of renal oxidative stress and injury were minimal or absent following infusion of Hb or NaNO2 alone. Together, these results suggest that the exposure to NaNO2 in settings associated with increased extracellular Hb may potentiate acute renal toxicity via processes that are independent of NaNO2 induced erythrocyte methemoglobinemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501713
Anti-NFE2L2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002990 Anti-NFE2L2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701989
Anti-NFE2L2 antibody produced in rabbit
SAB1406635
Anti-NPHS2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV47047
Anti-NPHS2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA049486
Anti-NPHS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411415
Anti-NPHS2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501984
Anti-NRF2, C-Terminal antibody produced in rabbit, affinity isolated antibody
P0372
Anti-Podocin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2102716
Anti-WT1 antibody produced in rabbit, affinity isolated antibody
SAB2104329
Anti-WT1 antibody produced in rabbit, affinity isolated antibody
SAB1411399
Anti-WT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0004780M1
Monoclonal Anti-NFE2L2 antibody produced in mouse, clone 1F3, purified immunoglobulin, buffered aqueous solution
SAB1404123
Monoclonal Anti-NFE2L2 antibody produced in mouse, clone 2G7, purified immunoglobulin, buffered aqueous solution
SAB1409360
Monoclonal Anti-NFE2L2 antibody produced in mouse, clone 3G7, purified immunoglobulin, buffered aqueous solution
SAB1409361
Monoclonal Anti-NFE2L2 antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution
SAB1409362
Monoclonal Anti-NFE2L2 antibody produced in mouse, clone 4C3, purified immunoglobulin, buffered aqueous solution
SAB1409363
Monoclonal Anti-NFE2L2 antibody produced in mouse, clone 1G10, purified immunoglobulin, buffered aqueous solution
WH0007490M1
Monoclonal Anti-WT1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1404530
Monoclonal Anti-WT1 antibody produced in mouse, clone 3B12, purified immunoglobulin, buffered aqueous solution
SAB1409794 Monoclonal Anti-WT1 antibody produced in mouse, clone 1A12, purified immunoglobulin, buffered aqueous solution
SAB1409795 Monoclonal Anti-WT1 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB1409796
Monoclonal Anti-WT1 antibody produced in mouse, clone 1E9, purified immunoglobulin, buffered aqueous solution