EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Proteome-wide identification and quantification of S-glutathionylation targets in mouse liver.
PMID 25891661
Abstract
Protein S-glutathionylation is a reversible post-translational modification regulating sulfhydryl homeostasis. However, little is known about the proteins and pathways regulated by S-glutathionylation in whole organisms and current approaches lack the sensitivity to examine this modification under basal conditions. We now report the quantification and identification of S-glutathionylated proteins from animal tissue, using a highly sensitive methodology combining high-accuracy proteomics with tandem mass tagging to provide precise, extensive coverage of S-glutathionylated targets in mouse liver. Critically, we show significant enrichment of S-glutathionylated mitochondrial and Krebs cycle proteins, identifying that S-glutathionylation is heavily involved in energy metabolism processes inxa0vivo. Furthermore, using mice nulled for GST Pi (GSTP) we address the potential for S-glutathionylation to be mediated enzymatically. The data demonstrate the impact of S-glutathionylation in cellular homeostasis, particularly in relation to energy regulation and is of significant interest for those wishing to examine S-glutathionylation in an animal model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV54290
Anti-ETFA (AB1) antibody produced in rabbit, affinity isolated antibody
AV54289
Anti-ETFA (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2501356
Anti-ETFA (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501355
Anti-ETFA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701648
Anti-etfa antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA018990
Anti-ETFA antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018993
Anti-ETFA antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018996
Anti-ETFA antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024089
Anti-ETFA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501231
Anti-TEAD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102402
Anti-TEAD2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2104126
Anti-TEAD2 antibody produced in rabbit, affinity isolated antibody
SAB4503373
Anti-TEAD2 antibody produced in rabbit, affinity isolated antibody
SAB1402767
Monoclonal Anti-ETFA antibody produced in mouse, clone 2A5, purified immunoglobulin, buffered aqueous solution
WH0008463M1 Monoclonal Anti-TEAD2 antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution