EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Decreased plasma levels of activated factor VII in patients with deep vein thrombosis.
PMID 25891834
Abstract
The initiating trigger in the development of deep vein thrombosis (DVT) remains unidentified. It has been suggested that tissue factor (TF)-bearing microparticles play a key role, which indicates a role for the TF pathway in the initiation of DVT. To assess the role of the TF pathway in the initiation of venous thrombosis, we measured plasma levels of factor VII and VIIa in patients with acute DVT and in controls. We included 148 patients diagnosed with acute DVT and 179 controls in this study. Antigen levels of FVII and FVIIa were measured by using assays recently developed in our laboratory. Median FVII levels in patients were 109.8% (interquartile range [IQR] 86.0-153.2) compared with 102.2% (IQR 76.1-141.7) in controls. Individuals with FVII levels in the upper quartile had a 1.6-fold increased risk for the presence of a DVT (odds ratio 1.6, 95% confidence interval 0.8-3.1). Median FVIIa levels in patients were 50.2xa0ngxa0mL(-1) (IQR 25.2-86.1) compared with 96.6xa0ngxa0mL(-1) (69.9-168.9) in controls. Individuals with FVIIa levels in the lowest quartile had a > 5-fold increased risk for the presence of a DVT (odds ratio 5.5, 95% confidence interval 2.8-10.6). Both risks did not change substantially after adjustment for potential confounders. Decreased plasma levels of FVIIa in patients with deep vein thrombosis may indicate ongoing consumption of FVIIa and suggest a contributory role for TF in venous thrombus formation.