EMAIL THIS PAGE TO A FRIEND
Developmental and comparative immunology
Molecular and functional characterization of porcine Siglec-3/CD33 and analysis of its expression in blood and tissues.
PMID 25892023
Abstract
A cDNA clone encoding a 380 a-a type 1 transmembrane protein with homology to human Siglec-3/CD33 was obtained from a swine small intestine library. An analysis of protein sequence identified two immunoglobulin-like domains, a transmembrane region, and a carboxi-terminal tail with two tyrosine-based signalling motifs. Binding assays of Siglec-3 transfected CHO cells to polyacrylamide glycoconjugates showed a preference for α2-6-linked sialic acids. Using mAbs raised against a fragment containing the two Ig-like domains, porcine Siglec-3 was found to be expressed on monocytes and granulocytes, and their bone marrow precursors. It was also detected in lymph node, splenic and alveolar macrophages. MAbs immunoprecipitated, from granulocyte lysates, a protein of 51-60 kDa under both non-reducing and reducing conditions. MAbs were also used to analyse functional activity of Siglec-3 on bone marrow and blood cells. Engagement of Siglec-3 by mAb had no apparent effect on cell proliferation or cytokine production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5159
Imiquimod, ≥98% (HPLC), solid
C14H16N4