EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Improvement in the blastocyst quality and efficiency of putative embryonic stem cell line derivation from porcine embryos produced in vitro using a novel culturing system.
PMID 25892608
Abstract
Porcine embryonic stem cells (pESCs) have great potential for application in translational biomedical research, including xenotransplantation and disease models. Obtaining high-quality blastocysts is the most important factor in the isolation and colonization of primary ESCs and the establishment of ESC lines. In pigs, in vitro-derived blastocysts have a limited cell number compared to in vivo-derived blastocysts and show an indefinite inner cell mass, which may result in failure to establish pESC lines. In the present study, the effects of resveratrol (RES), granulocyte-macrophage colony stimulating factor (GM-CSF) and β-mercaptoethanol (β-ME) on the quality of blastocysts and the efficiency of colony derivation were investigated for the establishment of ESCs. A novel culturing system was developed in which 2 µM RES was added to the oocyte in vitro maturation (IVM) medium, and 10 ng/ml pGM-CSF and 10 µM β-ME were added to embryo in vitro culture (IVC) medium. This novel system showed significantly more parthenogenetic activation (PA) blastocysts (54.5 ± 1.8% vs. 43.4 ± 1.2%; P<0.05) and in vitro fertilization (IVF) blastocysts (36.9 ± 3.3% vs. 26.2 ± 2.9%; P<0.06) at day seven as compared with that in the control system. The PA and IVF blastocysts from the novel system showed a significantly greater hatching rate (P<0.05) and greater cell numbers (55.1 ± 2.0 vs. 45.6 ± 2.0; P<0.05 and 78.9 ± 6.8 vs. 58.5 ± 7.2; P<0.06, for PA and IVF, respectively) at day seven compared to that in the control system. After seeding on feeder cells, the PA blastocysts produced by the novel system showed a significantly increased rate of attachment (28.8 ± 3.9% vs. 17.2 ± 2.4%; P<0.062). Finally, two putative pESC lines from PA embryos produced by the novel system and one by the control system were established. In conclusion, the novel system improved blastocyst quality and increased the derivation efficiency of putative pESC lines from porcine PA and IVF embryos produced in vitro.