EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Enantiomer separation of acidic chiral compounds on a quinine-silica/zirconia hybrid monolith by capillary electrochromatography.
PMID 25892638
Abstract
A weak anion-exchanger chiral selector, quinine-incorporated silica/zirconia hybrid monolithic (QUI-S/ZHM) capillary column was prepared by sol-gel technology. The performance of the QUI-S/ZHM column was investigated for enantioresolution of a set of acidic chiral drugs and dinitrobenzoyl (DNB)-amino acids by capillary electrochromatography in aqueous organic mobile phases composed of acetonitrile (ACN) and triethylammonium acetate (TEAA) buffer. Effects of several parameters including the ACN content, concentration and pH of the mobile phase on the chiral separation were examined. Baseline resolutions of all the compounds were obtained in the mobile phase consisting of 70:30 ACN/TEAA (10mM, pH 6) under applied voltage of -10kV at 25°C within 20min.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
413364
3-(Triethoxysilyl)propyl isocyanate, 95%
C10H21NO4Si
145904
Quinine, 90%
C20H24N2O2