EMAIL THIS PAGE TO A FRIEND
Nature communications
Extracellular MRP8/14 is a regulator of β2 integrin-dependent neutrophil slow rolling and adhesion.
PMID 25892652
Abstract
Myeloid-related proteins (MRPs) 8 and 14 are cytosolic proteins secreted from myeloid cells as proinflammatory mediators. Currently, the functional role of circulating extracellular MRP8/14 is unclear. Our present study identifies extracellular MRP8/14 as an autocrine player in the leukocyte adhesion cascade. We show that E-selectin-PSGL-1 interaction during neutrophil rolling triggers Mrp8/14 secretion. Released MRP8/14 in turn activates a TLR4-mediated, Rap1-GTPase-dependent pathway of rapid β2 integrin activation in neutrophils. This extracellular activation loop reduces leukocyte rolling velocity and stimulates adhesion. Thus, we identify Mrp8/14 and TLR4 as important modulators of the leukocyte recruitment cascade during inflammation in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2702075
Anti-AP3B1 (N-terminal) antibody produced in rabbit
SAB1409916
Anti-AP3B1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA038737 Anti-AP3B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106308
Anti-AP3B1 antibody produced in rabbit, affinity isolated antibody
SAB2501783 Anti-PREP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104775
Anti-PREP antibody produced in rabbit, affinity isolated antibody
HPA031388
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031389
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031390
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104515
Anti-PREP, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2500903
Anti-S100A9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA004193
Anti-S100A9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411220
Anti-S100A9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2104737
Anti-S100A9, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1412080
ANTI-TLR4 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
SAB1412548
ANTI-TLR4 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1412602
ANTI-TLR4 antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
PRS3141
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500765
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody
WH0008546M6
Monoclonal Anti-AP3B1 antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
SAB1401302
Monoclonal Anti-PREP antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution
SAB1404348
Monoclonal Anti-S100A9 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409623
Monoclonal Anti-S100A9 antibody produced in mouse, clone 1C22, purified immunoglobulin, buffered aqueous solution
SAB1404475
Monoclonal Anti-TLR4 antibody produced in mouse, clone 3B6, purified immunoglobulin, buffered aqueous solution
SAB1402375
Monoclonal Anti-TLR4 antibody produced in mouse, clone 1H7, purified immunoglobulin, buffered aqueous solution