EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Fermentation characteristics and angiotensin I-converting enzyme-inhibitory activity of Lactobacillus helveticus isolate H9 in cow milk, soy milk, and mare milk.
PMID 25892687
Abstract
Lactobacillus helveticus isolate H9 demonstrated high angiotensin I-converting enzyme (ACE)-inhibitory activity in previous research. Here, we evaluated the fermentation characteristics (pH, titratable acidity, free amino nitrogen, and viable bacterial counts), ACE-inhibitory activity, and contents of Val-Pro-Pro (VPP) and Ile-Pro-Pro (IPP) peptides of stored yogurt (4°C for 28 d) fermented by L. helveticus isolate H9 (initially inoculated at 4 concentrations), from cow, mare, and soy milks. During storage, the pH and titratable acidity remained stable in yogurts produced from all milk types and all inoculation concentrations. The viable bacterial counts in all stored yogurts ranged between 10(6.72) and 10(8.59) cfu/g. The highest ACE-inhibitory activity (70.9-74.5%) was achieved at inoculation concentrations of 5×10(6) cfu/mL. The ACE-inhibitory tripeptides VPP and IPP as determined by ultra-performance liquid chromatography-tandem mass spectrometry were not produced in yogurt made from soy milk or mare milk. These evaluations indicate that L. helveticus H9 has good probiotic properties and would be a promising candidate for production of fermented food with probiotic properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56395
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
C10H17NO3