EMAIL THIS PAGE TO A FRIEND
Spine
Does Magnetic Resonance Imaging Improve the Predictive Performance of a Validated Clinical Prediction Rule Developed to Evaluate Surgical Outcome in Patients With Degenerative Cervical Myelopathy?
PMID 25893357
Abstract
Ambispective study. To determine whether MRI parameters improve the predictive performance of a validated clinical prediction rule used to assess functional outcomes in surgical patients with DCM. Degenerative cervical myelopathy (DCM) is the most common cause of spinal cord dysfunction in the elderly worldwide. A clinical prediction rule was developed to discriminate between patients with mild myelopathy postoperatively (mJOA ≥ 16) and those with substantial residual neurological impairment (mJOA < 16). Recently, a separate magnetic resonance imaging (MRI)-based prediction model was created. However, a model exploring the combined predictive value of imaging and clinical variables does not exist. One hundred and fourteen patients with MRIs were examined from a cohort of 278 patients enrolled in the AOSpine CSM-North America Study. Ninety-nine patients had complete preoperative imaging and postoperative outcome data. MRIs were evaluated for the presence/absence of signal change on T2- and T1-weighted images. Quantitative analysis of the T2 signal change was conducted and maximum canal compromise and cord compression were calculated. The added predictive performance of each MRI parameter to the clinical model was evaluated using receiver operator characteristic curves. The model developed on our subsample yielded an area under the receiver operator curve (AUC) of 0.811 (95% CI: 0.726-0.896). The addition of imaging variables did not significantly improve the predictive performance. Small improvements in prediction were obtained when sagittal extent of T2 hyperintensity (AUC: 0.826, 95% CI: 0.743-0.908, 1.35% increase) or Wang ratio (AUC: 0.823, 95% CI: 0.739-0.907, 1.21%) was added. Anatomic characteristics, such as maximum canal compromise and maximum cord compression, did not improve the discriminative ability of the clinical prediction model. In our sample of surgical patients, with clinical and image-evidence of DCM, MRI parameters do not significantly add to the predictive performance of a previously published clinical prediction rule. It remains plausible that combinations of the strongest clinical and MRI predictors may yield a similar or a superior prediction model. 3.