EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Poly(A)-specific ribonuclease deficiency impacts telomere biology and causes dyskeratosis congenita.
PMID 25893599
Abstract
Dyskeratosis congenita (DC) and related syndromes are inherited, life-threatening bone marrow (BM) failure disorders, and approximately 40% of cases are currently uncharacterized at the genetic level. Here, using whole exome sequencing (WES), we have identified biallelic mutations in the gene encoding poly(A)-specific ribonuclease (PARN) in 3 families with individuals exhibiting severe DC. PARN is an extensively characterized exonuclease with deadenylation activity that controls mRNA stability in part and therefore regulates expression of a large number of genes. The DC-associated mutations identified affect key domains within the protein, and evaluation of patient cells revealed reduced deadenylation activity. This deadenylation deficiency caused an early DNA damage response in terms of nuclear p53 regulation, cell-cycle arrest, and reduced cell viability upon UV treatment. Individuals with biallelic PARN mutations and PARN-depleted cells exhibited reduced RNA levels for several key genes that are associated with telomere biology, specifically TERC, DKC1, RTEL1, and TERF1. Moreover, PARN-deficient cells also possessed critically short telomeres. Collectively, these results identify a role for PARN in telomere maintenance and demonstrate that it is a disease-causing gene in a subset of patients with severe DC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500313 Anti-DKC1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405716
Anti-DKC1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000166
Anti-DKC1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200611
Anti-DKC1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA000447
Anti-DKC1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104539
Anti-DKC1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4500657
Anti-Dyskerin antibody produced in rabbit, affinity isolated antibody
SAB4501766
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501767
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501764
Anti-Lamin A antibody produced in rabbit, affinity isolated antibody
SAB1406087
Anti-LMNA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006660
Anti-LMNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406220
Anti-PARN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006314
Anti-PARN antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37393
Anti-PARN antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
HPA012010
Anti-PARN antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502943
Anti-Telomeric Repeat Binding Factor 1 antibody produced in rabbit, affinity isolated antibody
E1521
Ethylenediamine, BioXtra
C2H8N2
391085
Ethylenediamine, purified by redistillation, ≥99.5%
C2H8N2
E26266
Ethylenediamine, ReagentPlus®, ≥99%
C2H8N2
E1649
Ethylenediamine, meets USP testing specifications
C2H8N2
15068
Ethylenediamine solution, technical, 75-80%
C2H8N2
EHUEGFP MISSION® esiRNA, esiRNA targeting EGFP (esiRNA1)
T1948
Monoclonal Anti-TRF1 antibody produced in mouse, ~2 mg/mL, clone TRF-78, purified immunoglobulin, buffered aqueous solution
T7567
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
C12H27P
335940
Xylene Cyanol FF, Dye content 75 %
C25H27N2NaO6S2
X4126
Xylene Cyanol FF, for molecular biology, BioReagent
C25H27N2NaO6S2