EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
mTORC1 and mTORC2 selectively regulate CD8⁺ T cell differentiation.
PMID 25893604
Abstract
Activation of mTOR-dependent pathways regulates the specification and differentiation of CD4+ T effector cell subsets. Herein, we show that mTOR complex 1 (mTORC1) and mTORC2 have distinct roles in the generation of CD8+ T cell effector and memory populations. Evaluation of mice with a T cell-specific deletion of the gene encoding the negative regulator of mTORC1, tuberous sclerosis complex 2 (TSC2), resulted in the generation of highly glycolytic and potent effector CD8+ T cells; however, due to constitutive mTORC1 activation, these cells retained a terminally differentiated effector phenotype and were incapable of transitioning into a memory state. In contrast, CD8+ T cells deficient in mTORC1 activity due to loss of RAS homolog enriched in brain (RHEB) failed to differentiate into effector cells but retained memory characteristics, such as surface marker expression, a lower metabolic rate, and increased longevity. However, these RHEB-deficient memory-like T cells failed to generate recall responses as the result of metabolic defects. While mTORC1 influenced CD8+ T cell effector responses, mTORC2 activity regulated CD8+ T cell memory. mTORC2 inhibition resulted in metabolic reprogramming, which enhanced the generation of CD8+ memory cells. Overall, these results define specific roles for mTORC1 and mTORC2 that link metabolism and CD8+ T cell effector and memory generation and suggest that these functions have the potential to be targeted for enhancing vaccine efficacy and antitumor immunity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2538
Anti-Hamartin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300187
Anti-phospho-RPS6 (pSer235) antibody produced in rabbit, affinity isolated antibody
HPA002044
Anti-PSMC4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005471
Anti-PSMC4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105008
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2105009
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300484
Anti-RPS6 (Ab-235) antibody produced in rabbit, affinity isolated antibody
SAB1300250
Anti-RPS6 (N-term) antibody produced in rabbit, Ig fraction of antiserum, buffered aqueous solution
HPA031153
Anti-RPS6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107629
Anti-RPS6 antibody produced in rabbit, affinity isolated antibody
PRS3503
Anti-TSC1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3505
Anti-TSC1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3507
Anti-TSC2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3509
Anti-TSC2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA030409
Anti-TSC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503037
Anti-TSC2 antibody produced in rabbit, affinity isolated antibody
SAB4503038
Anti-TSC2, C-Terminal antibody produced in rabbit, affinity isolated antibody
T9574
Anti-Tuberin (IA-22) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
T5075 Anti-Tuberin (VV-18) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503039
Anti-Tuberin antibody produced in rabbit, affinity isolated antibody
WH0005704M1
Monoclonal Anti-PSMC4 antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
WH0006194M1
Monoclonal Anti-RPS6 antibody produced in mouse, clone 3H1-F2, purified immunoglobulin, buffered aqueous solution
SAB1402949
Monoclonal Anti-TSC2 antibody produced in mouse, clone 1C1, purified immunoglobulin, buffered aqueous solution
PLA0118
Rabbit anti-Phospho RPS6 (S235/236) Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.