EMAIL THIS PAGE TO A FRIEND
Cell biology and toxicology
Hepatic differentiation of human adipose tissue-derived mesenchymal stem cells and adverse effects of arsanilic acid and acetaminophen during in vitro hepatic developmental stage.
PMID 25894252
Abstract
In the present study, we differentiated hepatocyte-like cells (HLCs) from human adipose tissue-derived mesenchymal stem cells (AT-MSCs). The hepatic differentiation was confirmed by increases in hepatic proteins or genes, the cytochrome P450 (CYP) activities, albumin secretion, and glycogen storage. To determine the developmental toxic effect of arsanilic acid (Ars) and acetaminophen (AAP) on the hepatic development, the differentiating cells were treated with the test chemicals (below IC12.5) from day 4 to day 13. The enzymatic activities of lactate dehydrogenase (LDH), alanine aminotransferase (ALT), and aspartate aminotransferase (AST) did not significantly differ in response to Ars treatment. AAP treatment increased the activities of all enzymes in a dose-dependent manner, significantly at concentrations of 2.5 and 5 mM of AAP. On the expressions of hepatic genes for Ars, the expressions were significantly inhibited by more than 0.5 mM for Albumin (ALB), but only 2.5 mM for α-feto protein (AFP). In the AAP-treated group, the expressions of ALB and AFP were significantly decreased at the concentrations exceeding 0.625 mM. The activities of CYP3A4 were not changed by both treatments. The activities of CYP1A2 were increased by AAP, whereas it was decreased by Ars treatment. In conclusion, AAP could cause serious adverse effects during the hepatic development as compared to Ars.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306569
ANTI-CYK18(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501665
Anti-Keratin 18, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300551
Anti-KRT18 (Ab-33) antibody produced in rabbit, affinity isolated antibody
AV40205
Anti-KRT18 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV40206
Anti-KRT18 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22753 Anti-KRT18 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2701928
Anti-krt18 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3300015
Monoclonal Anti-CK18 antibody produced in mouse, clone 10, purified immunoglobulin, buffered aqueous glycerol solution
SAB3300017 Monoclonal Anti-CK18 antibody produced in mouse, clone 1H10, purified immunoglobulin, buffered aqueous glycerol solution
SAB1305526
MONOCLONAL ANTI-CYK18 (C-TERMINAL) antibody produced in mouse, clone 695CT17.2.6, crude ascites, buffered aqueous solution
SAB1305562
MONOCLONAL ANTI-CYK18(C-TERMINAL) antibody produced in mouse, clone 695CT17.2.6, IgG fraction of antiserum, buffered aqueous solution
WH0003875M1
Monoclonal Anti-KRT18 antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution
SAB1404012
Monoclonal Anti-KRT18 antibody produced in mouse, clone 1G8, purified immunoglobulin, buffered aqueous solution
SAB4700411
Monoclonal Anti-KRT18 antibody produced in mouse, clone C-04, purified immunoglobulin, buffered aqueous solution
SAB4700670 Monoclonal Anti-KRT18 antibody produced in mouse, clone DC-10, purified immunoglobulin, buffered aqueous solution