EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Sub-minute method for simultaneous determination of aspartame, cyclamate, acesulfame-K and saccharin in food and pharmaceutical samples by capillary zone electrophoresis.
PMID 25895731
Abstract
This paper reports the development of a sub-minute separation method by capillary zone electrophoresis for the determination of aspartame, cyclamate, acesulfame-K and saccharin in food products and pharmaceutical samples. Separations were performed in a fused uncoated silica capillary with UV detection at 220nm. Samples and standards were injected hydrodynamically using the short-end injection procedure. The electrophoretic system was operated under constant voltage of -30kV. The background electrolyte was composed of 45mmolL(-1) 2-amino-2-(hydroxymethyl)-1,3-propanediol and 15mmolL(-1) benzoic acid at pH 8.4. The separation time for all analytes was less than 1min. Evaluation of analytical parameters of the method showed good linearity (r(2)>0.9972), limit of detection of 3.3-6.4mgL(-1), intermediate precision better than 9.75% (peak area of sample) and recovery in the range of 91-117%.