EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Preparation and characterization of quaternary ammonium chitosan hydrogel with significant antibacterial activity.
PMID 25895959
Abstract
Quaternary ammonium chitosan (HACC)/polyvinyl alcohol (PVA)/polyethylene oxide (PEO) hydrogels were prepared using gamma radiation. The chemical structure of the hydrogels was characterized using FT-IR. The results revealed that HACC, PVA and PEO were perfectly compatible and interacted via the hydrogen bonds. As revealed by SEM, scaffolds with a homogeneous interconnected pore structure were obtained after lyophilizing the hydrogels. The influence of different radiation doses and weight ratios on properties including gel content, swelling ability, water evaporation rate and mechanical properties were investigated. It indicated that the hydrogels had the good swelling ability, water evaporation rate and mechanical properties. In vitro antibacterial activity assessment, the hydrogels exhibited a pronounced inhibitory effect against two bacteria (Staphylococcus aureus and Escherichia coli). Therefore, the hydrogels showed a promising potential to be applied as wound dressing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29469
N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, ≥99.0% (CHN)
C14H26N3O · C7H7O3S