EMAIL THIS PAGE TO A FRIEND
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology
Evaluation of several phenotypic methods for the detection of carbapenemase-producing Pseudomonas aeruginosa.
PMID 25896858
Abstract
The purpose of this investigation was to compare several phenotypic methods, including combined disk tests (CDT) containing metallo-β-lactamase (MBL) inhibitors or cloxacillin, and the Carba NP test for the detection of carbapenemase-producing Pseudomonas aeruginosa (CPPA). A new CDT using imipenem (10 μg) ± cloxacillin 4,000 μg and the Carba NP test were evaluated to detect CPPA. In addition, four commercially available combined disks containing a carbapenem and ethylene-diamine-tetra-acetic acid (EDTA) or dipicolinic acid (DPA) as the inhibitor were tested in order to detect MBL-positive P. aeruginosa. All these phenotypic methods were evaluated on 188 imipenem non-susceptible P. aeruginosa (CPPA, n = 75) isolates divided into 26 well-characterized collection strains and 162 non-duplicate clinical isolates referred to the national reference laboratory in 2013. For the total of 188 isolates tested, CDT containing EDTA or DPA displayed high sensitivities (99%) and specificities (95%) for detecting MBL-producing isolates. CDT with cloxacillin showed a sensitivity and specificity of 97%/96% compared to 88%/99% for the Carba NP test in order to detect CPPA. For the 162 clinical isolates, CDT containing EDTA or DPA displayed a high negative predictive value (NPV) (99%) for detecting MBL-producing isolates. CDT with cloxacillin showed an NPV of 98%, compared to 95% for the Carba NP test in order to detect CPPA. In our setting, CDT associating imipenem ± EDTA or ± DPA performed best for the detection of MBL-producing P. aeruginosa. Imipenem/imipenem-cloxacillin test yielded good NPV to exclude the presence of MBL in imipenem non-susceptible isolates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21861
1,1′-Carbonyl-di-(1,2,4-triazole), technical, ≥90% (T)
C5H4N6O