EMAIL THIS PAGE TO A FRIEND
Nature communications
Modelling breast cancer requires identification and correction of a critical cell lineage-dependent transduction bias.
PMID 25896888
Abstract
Clinically relevant human culture models are essential for developing effective therapies and exploring the biology and etiology of human cancers. Current breast tumour models, such as those from oncogenically transformed primary breast cells, produce predominantly basal-like properties, whereas the more common phenotype expressed by the vast majority of breast tumours are luminal. Reasons for this puzzling, yet important phenomenon, are not understood. We show here that luminal epithelial cells are significantly more resistant to viral transduction than their myoepithelial counterparts. We suggest that this is a significant barrier to generating luminal cell lines and experimental tumours in vivo and to accurate interpretation of results. We show that the resistance is due to lower affinity of luminal cells for virus attachment, which can be overcome by pretreating cells--or virus--with neuraminidase. We present an analytical method for quantifying transductional differences between cell types and an optimized protocol for transducing unsorted primary human breast cells in context.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5884 α-L-Fucosidase from bovine kidney, ammonium sulfate suspension, ≥2.0 units/mg protein (biuret)
SAB4300659
Anti-CD10 antibody produced in rabbit, affinity isolated antibody
SAB4501931
Anti-CD227, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501932
Anti-CD227, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501561
Anti-ITGA6 (LIGHT CHAIN, CLEAVED-GLU942) antibody produced in rabbit, affinity isolated antibody
HPA012696
Anti-ITGA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA027582
Anti-ITGA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410719
Anti-ITGA6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501670
Anti-Keratin 19 antibody produced in rabbit, affinity isolated antibody
SAB1409273
Anti-KRT14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV15002 Anti-KRT14 antibody produced in rabbit, IgG fraction of antiserum
HPA023040
Anti-KRT14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410741
Anti-KRT14 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101302
Anti-KRT19 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101303
Anti-KRT19 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1400148
Anti-KRT19 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA002465 Anti-KRT19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400166
Anti-MME antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401246
Anti-MME antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101529
Anti-MUC1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101530
Anti-MUC1 (ab2) antibody produced in rabbit, affinity isolated antibody
AV41446
Anti-MUC1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2101531
Anti-MUC1 (ab3) antibody produced in rabbit, affinity isolated antibody
GW22681 Anti-MUC1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA004179
Anti-MUC1 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA007235
Anti-MUC1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008855
Anti-MUC1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501933
Anti-MUC1, C-Terminal antibody produced in rabbit, affinity isolated antibody
G1288 β-(1→3,4,6)-Galactosidase, positionally specific, recombinant, expressed in E. coli, buffered aqueous solution
H9268
Hexadimethrine bromide, ≥94% (titration)