EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Microcystin-RR exposure results in growth impairment by disrupting thyroid endocrine in zebrafish larvae.
PMID 25897773
Abstract
Recent studies have shown that cyanobacteria-derived microcystins (MCs) have the potential to disrupt endocrine systems. However, the effects of microcystin-RR (MC-RR) and their underlying mechanisms are poorly resolved in fish. In this study, MC-RR exposure through submersion caused serious developmental toxicity, such as growth delay and depressed heart rates in zebrafish larvae. We also detected decreased levels of thyroid hormones (THs), suggesting that MC-RR-triggered thyroid endocrine disruption might contribute to the growth impairment observed in developing zebrafish. To further our understanding of mechanisms of MC-RR-induced endocrine toxicity, quantitative real-time PCR (QPCR) analysis was performed on hypothalamic-pituitary-thyroid (HPT) axis related genes, i.e., corticotropin-releasing factor (CRF), thyroid-stimulating hormone (TSH), sodium/iodide symporter (NIS), thyroglobulin (TG), thyroid receptors (TRα and TRβ) and iodothyronine deiodinases (Dio1 and Dio2), of developing zebrafish embryos exposed to 0, 0.3, 1.0 or 3.0mgL(-1) MC-RR until 96h post-fertilization. Our results showed that transcription pattern of HPT axis related genes were greatly changed by MC-RR exposure, except TG gene. Furthermore, western blot was used to validate the results of gene expression. The results showed protein synthesis of TG was not affected, while that of NIS was significantly up-regulated, which are in accordance with gene expression. The overall results indicated that exposure to MC-RR can induce developmental toxicity, which might be associated with thyroid endocrine disruption in developing zebrafish larvae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220051
N-Iodosuccinimide, 95%
C4H4INO2