EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Rapamycin Suppresses Tumor Growth and Alters the Metabolic Phenotype in T-Cell Lymphoma.
PMID 25897830
Abstract
The mTOR pathway is a master regulator of cellular growth and metabolism. The biosynthetic and energetic demand of rapidly proliferating cells such as cancer cells is met by metabolic adaptations such as an increased glycolytic rate known as the Warburg effect. Herein, we characterize the anti-tumor effect of rapamycin in a mouse model of T-cell lymphoma and examine the metabolic effects in vitro. The murine T-cell lymphoma line, MBL2, and human cutaneous T-cell lymphoma (CTCL) lines, HH and Hut78, were used in syngeneic or standard NSG mouse models to demonstrate a marked suppression of tumor growth by rapamycin accompanied by inhibition of mTORC1/2. Analysis of the metabolic phenotype showed a substantial reduction in the glycolytic rate and glucose utilization in rapamycin-treated lymphoma cells. This was associated with reduced expression of glucose transporters and glycolytic enzymes in cultured cells and xenograft tumors. As a result of the decrease in glycolytic state, rapamycin-treated cells displayed reduced sensitivity to low-glucose conditions but continued to rely on mitochondrial oxidative phosphorylation (OXPHOS) with sensitivity to inhibition of OXPHOS. Taken together, we demonstrate that rapamycin suppresses growth of T-cell lymphoma tumors and leads to a reduction in aerobic glycolysis counteracting the Warburg effect of cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8375
2-Deoxy-D-glucose, ≥98% (GC), crystalline
C6H12O5
D6134
2-Deoxy-D-glucose, ≥99% (GC), crystalline
C6H12O5
D3179
2-Deoxy-D-glucose, BioXtra, ≥98%
C6H12O5
SAB4300444
Anti-RPS6KB1 (Ab-389) antibody produced in rabbit, affinity isolated antibody
SAB4300515
Anti-RPS6KB1 (Ab-411) antibody produced in rabbit, affinity isolated antibody
SAB4300500
Anti-RPS6KB1 (Ab-421) antibody produced in rabbit, affinity isolated antibody
SAB4300527
Anti-RPS6KB1 (Ab-424) antibody produced in rabbit, affinity isolated antibody
SAB1412617
ANTI-RPS6KB1 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB1412626
ANTI-RPS6KB1 antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution
SAB2104784
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2104785
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
S4047 Anti-S6 Kinase antibody produced in rabbit, whole antiserum
SAB4502691
Anti-S6K antibody produced in rabbit, affinity isolated antibody
SAB1404335
Monoclonal Anti-RPS6KB1, (C-terminal) antibody produced in mouse, clone 1E10, purified immunoglobulin, buffered aqueous solution
R8781 Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
C51H79NO13
R0395
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
C51H79NO13