EMAIL THIS PAGE TO A FRIEND
Journal of molecular neuroscience : MN
Death Receptor 6 and Caspase-6 Regulate Prion Peptide-Induced Axonal Degeneration in Rat Spinal Neurons.
PMID 25898930
Abstract
Axonal degeneration is a hallmark of many neurodegenerative disorders including transmissible spongiform encephalopathies (TSE). However, the full complement of axonal degeneration triggers is not fully understood. In an in vitro prion model, we observed that treatment of rat spinal neurons with the prion peptide, PrP106-126, activated death receptor 6 (DR6, also known as TNFRSF21), caspase-6, caspase-3, and induced axonal degeneration. Knockdown of DR6 by siRNA blocked caspase-6 and caspase-3 activation and axonal degeneration. We also found that cleaved caspase-3 is only enriched in cell bodies, but cleaved caspase-6 is expressed in both cell bodies and axons. Axonal degeneration was prevented by preincubation of neurons with a caspase-6 inhibitor or siRNA of caspase-6. Our findings suggest that both DR6 and caspase-6 play important roles in axonal degeneration and caspase-6 acts downstream of DR6. We also observed that nicotinamide nucleotide adenylyltransferase 1 protein (Nmnat1), which had been reported to protect neurons from degeneration, alleviated axonal degeneration without blocking caspase-6 activation, suggesting that Nmnat acts downstream or parallel to caspase-6 activation. Our results indicate that PrP106-126 triggered axonal degeneration of the spinal cord neurons, DR6 is a key regulator of axonal degeneration, and the signaling pathway of DR6/caspase-6 mediates axonal degeneration induced by the prion fragment. Our findings raise the hope of targeting the DR6 as a potential therapeutic strategy in prion-related neurodegenerative diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA011337
Anti-CASP6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024303
Anti-CASP6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503308
Anti-CASP6, C-Terminal antibody produced in rabbit, affinity isolated antibody
C7599 Anti-Caspase 6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500402
Anti-Caspase-6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
D1564
Anti-DR6 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500255
Anti-DR6 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1407346
Anti-TNFRSF21 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006746
Anti-TNFRSF21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102490 Anti-TNFRSF21 antibody produced in rabbit, affinity isolated antibody
SAB2502108
Anti-TUBB3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4300623
Anti-Tubulin β-III antibody produced in rabbit, affinity isolated antibody
WH0004744M1
Monoclonal Anti-NEFH antibody produced in mouse, clone 2E8, purified immunoglobulin, buffered aqueous solution
WH0027242M8
Monoclonal Anti-TNFRSF21 antibody produced in mouse, clone 1B2, purified immunoglobulin, buffered aqueous solution
SAB1405038
Monoclonal Anti-TNFRSF21 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB3300047 Monoclonal Anti-TUBB3 antibody produced in mouse, clone 5H2, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700544
Monoclonal Anti-TUBB3 antibody produced in mouse, clone TU-20, purified immunoglobulin, buffered aqueous solution