EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Approaching 'Kit-Type' Labelling with (68)Ga: The DATA Chelators.
PMID 25899500
Abstract
The DATA chelators are a novel class of tri-anionic ligands based on 6-amino-1,4-diazepine-triacetic acid, which have been introduced recently for the chelation of (68)Ga. Compared with macrocyclic chelators based on the cyclen scaffold (i.e., DOTA, DO3A, and DO2A derivatives), DATA chelators undergo quantitative radiolabelling more rapidly and under milder conditions. In this study, a systematic evaluation of the labelling of four DATA chelators--DATA(M), DATA(P), DATA(Ph), and DATA(PPh)--with (68)Ga is presented. The results highlight the extraordinary potential of this new class of chelators for application in molecular imaging using (68)Ga positron emission tomography (PET).