EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Bioactive glass ions as strong enhancers of osteogenic differentiation in human adipose stem cells.
PMID 25900445
Abstract
Bioactive glasses are known for their ability to induce osteogenic differentiation of stem cells. To elucidate the mechanism of the osteoinductivity in more detail, we studied whether ionic extracts prepared from a commercial glass S53P4 and from three experimental glasses (2-06, 1-06 and 3-06) are alone sufficient to induce osteogenic differentiation of human adipose stem cells. Cells were cultured using basic medium or osteogenic medium as extract basis. Our results indicate that cells stay viable in all the glass extracts for the whole culturing period, 14 days. At 14 days the mineralization in osteogenic medium extracts was excessive compared to the control. Parallel to the increased mineralization we observed a decrease in the cell amount. Raman and Laser Induced Breakdown Spectroscopy analyses confirmed that the mineral consisted of calcium phosphates. Consistently, the osteogenic medium extracts also increased osteocalcin production and collagen Type-I accumulation in the extracellular matrix at 13 days. Of the four osteogenic medium extracts, 2-06 and 3-06 induced the best responses of osteogenesis. However, regardless of the enhanced mineral formation, alkaline phosphatase activity was not promoted by the extracts. The osteogenic medium extracts could potentially provide a fast and effective way to differentiate human adipose stem cells in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409631
Anti-BGLAP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1405515
Anti-CEACAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C4830
Calcium carbonate, BioXtra, ≥99.0%
CCaO3
C5929
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
CCaO3
481807
Calcium carbonate, 99.999% trace metals basis
CCaO3
202932
Calcium carbonate, ≥99.995% trace metals basis
CCaO3
310034
Calcium carbonate, powder, ≤30 μm particle size, 98%
CCaO3
21061
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
CCaO3
C0732
Cetylpyridinium chloride, meets USP testing specifications
C21H38ClN · H2O
WH0000632M1
Monoclonal Anti-BGLAP antibody produced in mouse, clone 2D4, purified immunoglobulin, buffered aqueous solution
SAB1401030
Monoclonal Anti-BGLAP antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution
WH0000634M1
Monoclonal Anti-CEACAM1 antibody produced in mouse, clone 2F10, purified immunoglobulin, buffered aqueous solution
SAB1403604
Monoclonal Anti-CEACAM1 antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
P1472
Potassium carbonate, meets USP testing specifications
CK2O3
590681
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
CK2O3
60108
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
CK2O3