EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The Evolutionarily Conserved C-terminal Domains in the Mammalian Retinoblastoma Tumor Suppressor Family Serve as Dual Regulators of Protein Stability and Transcriptional Potency.
PMID 25903125
Abstract
The retinoblastoma (RB) tumor suppressor and related family of proteins play critical roles in development through their regulation of genes involved in cell fate. Multiple regulatory pathways impact RB function, including the ubiquitin-proteasome system with deregulated RB destruction frequently associated with pathogenesis. With the current study we explored the mechanisms connecting proteasome-mediated turnover of the RB family to the regulation of repressor activity. We find that steady state levels of all RB family members, RB, p107, and p130, were diminished during embryonic stem cell differentiation concomitant with their target gene acquisition. Proteasome-dependent turnover of the RB family is mediated by distinct and autonomously acting instability elements (IE) located in their C-terminal regulatory domains in a process that is sensitive to cyclin-dependent kinase (CDK4) perturbation. The IE regions include motifs that contribute to E2F-DP transcription factor interaction, and consistently, p107 and p130 repressor potency was reduced by IE deletion. The juxtaposition of degron sequences and E2F interaction motifs appears to be a conserved feature across the RB family, suggesting the potential for repressor ubiquitination and specific target gene regulation. These findings establish a mechanistic link between regulation of RB family repressor potency and the ubiquitin-proteasome system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5037 Anti-Neurabin II (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
N5162 Anti-Neurabin II antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1303872 ANTI-PLCL1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA031849
Anti-PLCL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306820
ANTI-PPP1R9B(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500914
Anti-RAB3GAP1 antibody produced in rabbit, affinity isolated antibody
SAB4300387
Anti-RB1 (Ab-780) antibody produced in rabbit, affinity isolated antibody
SAB4300385
Anti-RB1 (Ab-795) antibody produced in rabbit, affinity isolated antibody
SAB4300386
Anti-RB1 (Ab-807) antibody produced in rabbit, affinity isolated antibody
AV33212
Anti-RB1 antibody produced in rabbit, affinity isolated antibody
SAB2102983
Anti-RBL1 antibody produced in rabbit, affinity isolated antibody
HPA054962
Anti-RBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA056525
Anti-RBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502595
Anti-RBL1 antibody produced in rabbit, affinity isolated antibody
SAB2104003
Anti-RBL1, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA019703
Anti-RBL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502597
Anti-RBL2, C-Terminal antibody produced in rabbit, affinity isolated antibody
P4360 Monoclonal Anti-p107 antibody produced in mouse, clone KAB6, purified immunoglobulin, buffered aqueous solution
P4485 Monoclonal Anti-p130 antibody produced in mouse, clone KAB40, purified immunoglobulin, buffered aqueous solution
SAB5300213 Monoclonal Anti-RB1 antibody produced in mouse, clone 7E4B8, ascites fluid
WH0005933M1
Monoclonal Anti-RBL1 antibody produced in mouse, clone 1A5, purified immunoglobulin, buffered aqueous solution
SAB1409597
Monoclonal Anti-RBL2 antibody produced in mouse, clone 4D7, purified immunoglobulin, buffered aqueous solution
SAB1409598
Monoclonal Anti-RBL2 antibody produced in mouse, clone 2H7, purified immunoglobulin, buffered aqueous solution
SAB1409599 Monoclonal Anti-RBL2 antibody produced in mouse, clone 1F11, ascites fluid, solution
PZ0199
PD 0332991 isethionate, ≥98% (HPLC)
C24H29N7O2 · C2H6O4S
SRP0256
RB Active human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)