EMAIL THIS PAGE TO A FRIEND
Pediatric radiology
3-D cardiac MRI in free-breathing newborns and infants: when is respiratory gating necessary?
PMID 25903840
Abstract
Newborns and small infants have shallow breathing. To suggest criteria for when respiratory gating is necessary during cardiac MRI in newborns and infants. One-hundred ten data sets of newborns and infants with (n = 92, mean age: 1.9 ± 1.7 [SD] years) and without (n = 18, mean age: 1.6 ± 1.8 [SD] years) navigator gating were analysed retrospectively. The respiratory motion of the right hemidiaphragm was recorded and correlated to age, weight, body surface area and qualitative image quality on a 4-point score. Quantitative image quality assessment was performed (sharpness of the delineation of the ventricular septal wall) as well as a matched-pair comparison between navigator-gated and non-gated data sets. No significant differences were found in overall image quality or in the sharpness of the ventricular septal wall between gated and non-gated scans. A navigator acceptance of >80% was frequently found in patients ages <12 months, body surface area <0.40 m(2), body weight <10 kg and a size of <80 cm. Sequences without respiratory gating may be used in newborns and small infants, in particular if age <12 months, body surface area <0.40 m(2), body weight <10 kg and height <80 cm.