EMAIL THIS PAGE TO A FRIEND
FEBS letters
TRIM35 negatively regulates TLR7- and TLR9-mediated type I interferon production by targeting IRF7.
PMID 25907537
Abstract
Toll-like receptor 7 (TLR7) and TLR9 sense viral nucleic acids and induce type I IFN production, which must be properly controlled to avoid autoimmune diseases. Here, we report the negative regulation of TLR7/9-mediated type I IFN production by TRIM35. TRIM35 expression is induced by TLR7/9 stimulation and then interacts with IRF7, which is the master regulator of type I IFN response. Furthermore, TRIM35 promotes the K48-linked ubiquitination of IRF7 and induces its degradation via a proteasome-dependent pathway. Therefore, TRIM35 is a negative feedback regulator of TLR7/9-mediated type I IFN production due to its ability to suppress the stability of IRF7.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501565
Anti-IRF-3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406043
Anti-IRF3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004895
Anti-IRF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV31992
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV38061
Anti-IRF3 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB3500280
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501563
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501564
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV34872
Anti-TRIM35 (AB1) antibody produced in rabbit, affinity isolated antibody
AV39165
Anti-TRIM35 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501694
Anti-TRIM35 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA019647
Anti-TRIM35 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102552
Anti-TRIM35 antibody produced in rabbit, affinity isolated antibody
SAB2103161
Anti-TRIM35 antibody produced in rabbit, affinity isolated antibody
SAB4100036
Monoclonal Anti-IRF3 antibody produced in mouse, culture supernatant
SAB4100037
Monoclonal Anti-IRF3 antibody produced in mouse, culture supernatant
SAB1402824
Monoclonal Anti-IRF3 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
WH0023087M2
Monoclonal Anti-TRIM35 antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution