EMAIL THIS PAGE TO A FRIEND
Stroke
Resveratrol Preconditioning Protects Against Cerebral Ischemic Injury via Nuclear Erythroid 2-Related Factor 2.
PMID 25908459
Abstract
Nuclear erythroid 2 related factor 2 (Nrf2) is an astrocyte-enriched transcription factor that has previously been shown to upregulate cellular antioxidant systems in response to ischemia. Although resveratrol preconditioning (RPC) has emerged as a potential neuroprotective therapy, the involvement of Nrf2 in RPC-induced neuroprotection and mitochondrial reactive oxygen species production after cerebral ischemia remains unclear. The goal of our study was to study the contribution of Nrf2 to RPC and its effects on mitochondrial function. We used rodent astrocyte cultures and an in vivo stroke model with RPC. An Nrf2 DNA binding ELISA and protein analysis via Western blotting of downstream Nrf2 targets were performed to determine RPC-induced activation of Nrf2 in rat and mouse astrocytes. After RPC, mitochondrial function was determined by measuring reactive oxygen species production and mitochondrial respiration in both wild-type and Nrf2-/- mice. Infarct volume was measured to determine neuroprotection, whereas protein levels were measured by immunoblotting. We report that Nrf2 is activated by RPC in rodent astrocyte cultures, and that loss of Nrf2 reduced RPC-mediated neuroprotection in a mouse model of focal cerebral ischemia. In addition, we observed that wild-type and Nrf2-/- cortical mitochondria exhibited increased uncoupling and reactive oxygen species production after RPC treatments. Finally, Nrf2-/- astrocytes exhibited decreased mitochondrial antioxidant expression and were unable to upregulate cellular antioxidants after RPC treatment. Nrf2 contributes to RPC-induced neuroprotection through maintaining mitochondrial coupling and antioxidant protein expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF99005193
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF90559794
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF31519975
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF60677646
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF64815239
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF41418378
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF33640860
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF85367605
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF87981564
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF53022833
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF71274258
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF70055524
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF86610539
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF00832081
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF61911288
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF94645233
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF05710430
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF27632055
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF34573697
Manganese, rod, 35mm, diameter 2.0mm, cast, 99.5%
Mn
GF38495885
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mn
GF42039918
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF15798730
Manganese, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn