EMAIL THIS PAGE TO A FRIEND
Experimental biology and medicine (Maywood, N.J.)
Expression and promoter methylation status of hMLH1, MGMT, APC, and CDH1 genes in patients with colon adenocarcinoma.
PMID 25908636
Abstract
Colorectal cancer (CRC) is the third most common cancer in men and the second in women worldwide. CRC development is the result of genetic and epigenetic alterations accumulation in the epithelial cells of colon mucosa. In the present study, DNA methylation, an epigenetic event, was evaluated in tumoral and matching normal epithelium in a cohort of 61 CRC patients. The results confirmed and expanded knowledge for the tumor suppressor genes hMLH1, MGMT, APC, and CDH1. Promoter methylation was observed for all the examined genes in different percentage. A total of 71% and 10% of the examined cases were found to be methylated in two or more and in all genes, respectively. mRNA and protein levels were also evaluated. Promoter methylation of hMLH1, MGMT, APC, and CDH1 genes was present at the early stages of tumor's formation and it could also be detected in the normal mucosa. Correlations of the methylated genes with patient's age and tumor's clinicopathological characteristics were also observed. Our findings suggest that DNA methylation is a useful marker for tumor progression monitoring and that promoter methylation in certain genes is associated with more advanced tumor stage, poor differentiation, and metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA043536
Anti-FZR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106927
Anti-FZR1 antibody produced in rabbit, affinity isolated antibody
SAB2105778
Anti-FZR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW21975F Anti-MGMT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406122
Anti-MGMT antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2105451
Anti-MGMT antibody produced in rabbit, affinity isolated antibody
M8445
Anti-MLH1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
M8320
Anti-MLH1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4501881
Anti-MLH1 antibody produced in rabbit, affinity isolated antibody
SAB2104053
Anti-MLH1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1402580
Monoclonal Anti-FZR1 antibody produced in mouse, clone 4C4, purified immunoglobulin, buffered aqueous solution
SAB1405091
Monoclonal Anti-FZR1, (N-terminal) antibody produced in mouse, clone 3E12, purified immunoglobulin, buffered aqueous solution
M3068 Monoclonal Anti-MGMT antibody produced in mouse, clone MGMT-214, purified immunoglobulin, buffered aqueous solution
WH0004292M2
Monoclonal Anti-MLH1 antibody produced in mouse, clone M1, purified immunoglobulin, buffered aqueous solution
SAB5300249 Monoclonal Anti-MLH1 antibody produced in mouse, clone 4C9C7, ascites fluid
PLA0004
Rabbit anti-MLH1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.