EMAIL THIS PAGE TO A FRIEND
Current oncology (Toronto, Ont.)
Establishing a clinic-based pancreatic cancer and periampullary tumour research registry in Quebec.
PMID 25908910
Abstract
Enrolling patients in studies of pancreatic ductal adenocarcinoma (pdac) is challenging because of the high fatality of the disease. We hypothesized that a prospective clinic-based study with rapid ascertainment would result in high participation rates. Using that strategy, we established the Quebec Pancreas Cancer Study (qpcs) to investigate the genetics and causes of pdac and other periampullary tumours (pats) that are also rare and underrepresented in research studies. Patients diagnosed with pdac or pat were introduced to the study at their initial clinical encounter, with a strategy to enrol participants within 2 weeks of diagnosis. Patient self-referrals and referrals of unaffected individuals with an increased risk of pdac were also accepted. Family histories, epidemiologic and clinical data, and biospecimens were collected. Additional relatives were enrolled in families at increased genetic risk. The first 346 completed referrals led to 306 probands being enrolled, including 190 probands affected with pdac, who represent the population focus of the qpcs. Participation rates were 88.4% for all referrals and 89.2% for pdac referrals. Family history, epidemiologic and clinical data, and biospecimens were ascertained from 91.9%, 54.6%, and 97.5% respectively of patients with pdac. Although demographics and trends in risk factors in our patients were consistent with published statistics for patients with pdac, the qpcs is enriched for families with French-Canadian ancestry (37.4%), a population with recurrent germ-line mutations in hereditary diseases. Using rapid ascertainment, a pdac and pat research registry with high participation rates can be established. The qpcs is a valuable research resource and its enrichment with patients of French-Canadian ancestry provides a unique opportunity for studies of heredity in these diseases.